Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63590113-63590376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMEB2
Entrez Gene ID
26205
Gene Name
glucocorticoid modulatory element binding protein 2
Gene Aliases
GMEB-2, P79PIF, PIF79
UniGene
Hs.473286
Gene Chromosome Location
Chr.20: 63587594-63627101 on Build GRCh38
Transcripts
NM_012384.4 XM_005260202.4 XM_006723776.2 XM_011528779.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5366046 c.1524C>T p.A508A Chr.20: 63590158-63590158 on Build GRCh38
1615870 c.1456A>T p.T486S Chr.20: 63590226-63590226 on Build GRCh38
4100445 c.1436C>T p.T479M Chr.20: 63590246-63590246 on Build GRCh38
4934169 c.1390G>C p.D464H Chr.20: 63590292-63590292 on Build GRCh38
1615871 c.1433T>G p.L478R Chr.20: 63590249-63590249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776758045 Chr.20: 63590169-63590169 on Build GRCh38
rs779894285 Chr.20: 63590245-63590245 on Build GRCh38
rs771330766 Chr.20: 63590155-63590155 on Build GRCh38
rs556560125 Chr.20: 63590317-63590317 on Build GRCh38
rs1004482861 Chr.20: 63590131-63590131 on Build GRCh38
rs779162396 Chr.20: 63590236-63590236 on Build GRCh38
rs995028251 Chr.20: 63590317-63590319 on Build GRCh38
rs755167056 Chr.20: 63590191-63590191 on Build GRCh38
rs759958259 Chr.20: 63590289-63590289 on Build GRCh38
rs768215209 Chr.20: 63590320-63590320 on Build GRCh38
rs767585227 Chr.20: 63590200-63590200 on Build GRCh38
rs771556140 Chr.20: 63590158-63590158 on Build GRCh38
rs760652797 Chr.20: 63590163-63590163 on Build GRCh38
rs754288240 Chr.20: 63590301-63590301 on Build GRCh38
rs34205217 Chr.20: 63590299-63590299 on Build GRCh38
rs145516314 Chr.20: 63590138-63590138 on Build GRCh38
rs772904791 Chr.20: 63590269-63590269 on Build GRCh38
rs748595394 Chr.20: 63590144-63590144 on Build GRCh38
rs311496 Chr.20: 63590257-63590257 on Build GRCh38
rs752805008 Chr.20: 63590201-63590201 on Build GRCh38
rs538984558 Chr.20: 63590335-63590335 on Build GRCh38
rs754427496 Chr.20: 63590282-63590282 on Build GRCh38
rs746495579 Chr.20: 63590247-63590247 on Build GRCh38
rs967372533 Chr.20: 63590224-63590224 on Build GRCh38
rs761845041 Chr.20: 63590259-63590259 on Build GRCh38
rs201453334 Chr.20: 63590353-63590353 on Build GRCh38
rs745479741 Chr.20: 63590237-63590237 on Build GRCh38
rs981055401 Chr.20: 63590338-63590338 on Build GRCh38
rs749849967 Chr.20: 63590229-63590229 on Build GRCh38
rs769649568 Chr.20: 63590332-63590332 on Build GRCh38
rs913460866 Chr.20: 63590223-63590223 on Build GRCh38
rs898807447 Chr.20: 63590330-63590330 on Build GRCh38
rs751736161 Chr.20: 63590181-63590181 on Build GRCh38
rs140964265 Chr.20: 63590175-63590175 on Build GRCh38
rs748335338 Chr.20: 63590226-63590226 on Build GRCh38
rs148937299 Chr.20: 63590315-63590315 on Build GRCh38
rs184763784 Chr.20: 63590298-63590298 on Build GRCh38
rs778229317 Chr.20: 63590228-63590228 on Build GRCh38
rs200453223 Chr.20: 63590260-63590260 on Build GRCh38
rs764092751 Chr.20: 63590165-63590165 on Build GRCh38
rs750726028 Chr.20: 63590306-63590306 on Build GRCh38
rs771657948 Chr.20: 63590239-63590239 on Build GRCh38
rs752962412 Chr.20: 63590285-63590285 on Build GRCh38
rs774036963 Chr.20: 63590157-63590157 on Build GRCh38
rs551655648 Chr.20: 63590160-63590160 on Build GRCh38
rs779300596 Chr.20: 63590305-63590305 on Build GRCh38
rs748067783 Chr.20: 63590331-63590331 on Build GRCh38
rs757579300 Chr.20: 63590230-63590230 on Build GRCh38
rs762630644 Chr.20: 63590341-63590341 on Build GRCh38
rs765009548 Chr.20: 63590178-63590178 on Build GRCh38
rs768341716 Chr.20: 63590248-63590248 on Build GRCh38
rs1009291504 Chr.20: 63590284-63590284 on Build GRCh38
rs370420235 Chr.20: 63590302-63590302 on Build GRCh38
rs756634351 Chr.20: 63590146-63590146 on Build GRCh38
rs566776788 Chr.20: 63590214-63590214 on Build GRCh38
rs573487608 Chr.20: 63590290-63590290 on Build GRCh38
rs778068236 Chr.20: 63590149-63590149 on Build GRCh38
rs140213772 Chr.20: 63590137-63590137 on Build GRCh38
rs144783295 Chr.20: 63590329-63590329 on Build GRCh38
rs963193624 Chr.20: 63590141-63590141 on Build GRCh38
rs201863145 Chr.20: 63590154-63590154 on Build GRCh38

View Full Product Details