Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32275540-32275769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCK
Entrez Gene ID
3932
Gene Name
LCK proto-oncogene, Src family tyrosine kinase
Gene Aliases
IMD22, LSK, YT16, p56lck, pp58lck
UniGene
Hs.470627
Gene Chromosome Location
Chr.1: 32251239-32286167 on Build GRCh38
Transcripts
NM_001042771.2 NM_005356.4 XM_011541453.2 XM_011541454.2 XM_011541455.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6398967 c.413A>G p.E138G Chr.1: 32275604-32275604 on Build GRCh38
5984165 c.442C>G p.H148D Chr.1: 32275633-32275633 on Build GRCh38
5659850 c.481G>A p.G161R Chr.1: 32275672-32275672 on Build GRCh38
5984164 c.616C>G p.H206D Chr.1: 32275633-32275633 on Build GRCh38
6398966 c.587A>G p.E196G Chr.1: 32275604-32275604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911057675 Chr.1: 32275670-32275670 on Build GRCh38
rs774750034 Chr.1: 32275599-32275599 on Build GRCh38
rs746182093 Chr.1: 32275602-32275602 on Build GRCh38
rs775231665 Chr.1: 32275695-32275695 on Build GRCh38
rs11576030 Chr.1: 32275558-32275558 on Build GRCh38
rs775789532 Chr.1: 32275614-32275614 on Build GRCh38
rs1015545475 Chr.1: 32275634-32275634 on Build GRCh38
rs764942817 Chr.1: 32275662-32275662 on Build GRCh38
rs3738004 Chr.1: 32275728-32275728 on Build GRCh38
rs769580260 Chr.1: 32275621-32275621 on Build GRCh38
rs866092612 Chr.1: 32275624-32275624 on Build GRCh38
rs761524121 Chr.1: 32275656-32275656 on Build GRCh38
rs746980721 Chr.1: 32275556-32275556 on Build GRCh38
rs772512580 Chr.1: 32275607-32275607 on Build GRCh38
rs761327722 Chr.1: 32275631-32275631 on Build GRCh38
rs775454062 Chr.1: 32275559-32275559 on Build GRCh38
rs868644258 Chr.1: 32275664-32275664 on Build GRCh38
rs895734776 Chr.1: 32275671-32275671 on Build GRCh38
rs758071462 Chr.1: 32275718-32275718 on Build GRCh38
rs919808803 Chr.1: 32275713-32275713 on Build GRCh38
rs1014124218 Chr.1: 32275691-32275691 on Build GRCh38
rs868401007 Chr.1: 32275619-32275619 on Build GRCh38
rs969834765 Chr.1: 32275727-32275727 on Build GRCh38
rs771193758 Chr.1: 32275560-32275560 on Build GRCh38
rs11576031 Chr.1: 32275605-32275605 on Build GRCh38
rs866632097 Chr.1: 32275589-32275589 on Build GRCh38
rs763681220 Chr.1: 32275635-32275635 on Build GRCh38
rs974402364 Chr.1: 32275706-32275706 on Build GRCh38
rs756121108 Chr.1: 32275557-32275557 on Build GRCh38
rs560048294 Chr.1: 32275692-32275692 on Build GRCh38

View Full Product Details