Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 69295512-69295777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYEOV
Entrez Gene ID
26579
Gene Name
myeloma overexpressed
Gene Aliases
OCIM
UniGene
Hs.523848
Gene Chromosome Location
Chr.11: 69294138-69297287 on Build GRCh38
Transcripts
NM_001293291.1 NM_001293294.1 NM_001293296.1 NM_001300923.1 NM_001300924.1 NM_138768.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4652053 c.267G>A p.A89A Chr.11: 69295717-69295717 on Build GRCh38
6194358 c.264T>C p.V88V Chr.11: 69295714-69295714 on Build GRCh38
253774 c.174C>T p.L58L Chr.11: 69295624-69295624 on Build GRCh38
7378256 c.236G>A p.R79H Chr.11: 69295686-69295686 on Build GRCh38
931365 c.168G>A p.S56S Chr.11: 69295618-69295618 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776120285 Chr.11: 69295668-69295668 on Build GRCh38
rs760955057 Chr.11: 69295551-69295551 on Build GRCh38
rs375533191 Chr.11: 69295618-69295618 on Build GRCh38
rs757992357 Chr.11: 69295595-69295595 on Build GRCh38
rs754900328 Chr.11: 69295696-69295696 on Build GRCh38
rs763338972 Chr.11: 69295725-69295725 on Build GRCh38
rs761320177 Chr.11: 69295736-69295736 on Build GRCh38
rs1044498202 Chr.11: 69295604-69295605 on Build GRCh38
rs7117236 Chr.11: 69295669-69295669 on Build GRCh38
rs1007368614 Chr.11: 69295663-69295663 on Build GRCh38
rs375012773 Chr.11: 69295635-69295635 on Build GRCh38
rs775436449 Chr.11: 69295586-69295586 on Build GRCh38
rs115294924 Chr.11: 69295716-69295716 on Build GRCh38
rs374994116 Chr.11: 69295536-69295536 on Build GRCh38
rs758816213 Chr.11: 69295633-69295634 on Build GRCh38
rs761264687 Chr.11: 69295680-69295680 on Build GRCh38
rs553846326 Chr.11: 69295694-69295694 on Build GRCh38
rs117131950 Chr.11: 69295686-69295686 on Build GRCh38
rs376860566 Chr.11: 69295561-69295561 on Build GRCh38
rs139486495 Chr.11: 69295651-69295651 on Build GRCh38
rs774782477 Chr.11: 69295735-69295735 on Build GRCh38
rs772261259 Chr.11: 69295687-69295687 on Build GRCh38
rs181508661 Chr.11: 69295575-69295575 on Build GRCh38
rs765513428 Chr.11: 69295662-69295662 on Build GRCh38
rs370559525 Chr.11: 69295634-69295634 on Build GRCh38
rs775689189 Chr.11: 69295720-69295720 on Build GRCh38
rs751264879 Chr.11: 69295602-69295602 on Build GRCh38
rs769066266 Chr.11: 69295734-69295734 on Build GRCh38
rs756874600 Chr.11: 69295611-69295611 on Build GRCh38
rs767014133 Chr.11: 69295745-69295745 on Build GRCh38
rs750778261 Chr.11: 69295597-69295597 on Build GRCh38
rs550325881 Chr.11: 69295574-69295574 on Build GRCh38
rs780588702 Chr.11: 69295635-69295644 on Build GRCh38
rs570576931 Chr.11: 69295657-69295657 on Build GRCh38
rs780783710 Chr.11: 69295612-69295612 on Build GRCh38
rs201337397 Chr.11: 69295639-69295639 on Build GRCh38
rs367657249 Chr.11: 69295652-69295652 on Build GRCh38
rs747510565 Chr.11: 69295666-69295674 on Build GRCh38
rs778942272 Chr.11: 69295703-69295703 on Build GRCh38
rs568014391 Chr.11: 69295685-69295685 on Build GRCh38
rs186636295 Chr.11: 69295632-69295632 on Build GRCh38
rs372605188 Chr.11: 69295558-69295558 on Build GRCh38
rs200342464 Chr.11: 69295753-69295753 on Build GRCh38
rs371272001 Chr.11: 69295670-69295670 on Build GRCh38
rs745685581 Chr.11: 69295617-69295617 on Build GRCh38
rs763496982 Chr.11: 69295599-69295599 on Build GRCh38
rs200120334 Chr.11: 69295710-69295710 on Build GRCh38
rs938137093 Chr.11: 69295717-69295717 on Build GRCh38
rs1013563666 Chr.11: 69295600-69295600 on Build GRCh38
rs947919631 Chr.11: 69295727-69295727 on Build GRCh38
rs566364607 Chr.11: 69295708-69295708 on Build GRCh38
rs747043106 Chr.11: 69295644-69295644 on Build GRCh38
rs375123114 Chr.11: 69295751-69295751 on Build GRCh38
rs1014779612 Chr.11: 69295604-69295604 on Build GRCh38
rs142853719 Chr.11: 69295679-69295679 on Build GRCh38
rs866887884 Chr.11: 69295709-69295709 on Build GRCh38
rs1005510777 Chr.11: 69295613-69295613 on Build GRCh38
rs201404318 Chr.11: 69295549-69295549 on Build GRCh38
rs369184711 Chr.11: 69295546-69295546 on Build GRCh38
rs759927054 Chr.11: 69295570-69295570 on Build GRCh38
rs375530923 Chr.11: 69295704-69295704 on Build GRCh38
rs917713081 Chr.11: 69295719-69295719 on Build GRCh38

View Full Product Details