Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23778633-23778906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PITHD1
Entrez Gene ID
57095
Gene Name
PITH domain containing 1
Gene Aliases
C1orf128, HT014, TXNL1CL
UniGene
Hs.31819
Gene Chromosome Location
Chr.1: 23778386-23788232 on Build GRCh38
Transcripts
NM_020362.4 XM_011541804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7276151 c.146G>A p.G49D Chr.1: 23778661-23778661 on Build GRCh38
5694543 c.144G>A p.E48E Chr.1: 23778659-23778659 on Build GRCh38
5511474 c.134A>G p.E45G Chr.1: 23778649-23778649 on Build GRCh38
6431633 c.157G>T p.G53C Chr.1: 23778672-23778672 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs990107369 Chr.1: 23778847-23778847 on Build GRCh38
rs982755468 Chr.1: 23778750-23778750 on Build GRCh38
rs914479588 Chr.1: 23778878-23778878 on Build GRCh38
rs897756695 Chr.1: 23778766-23778766 on Build GRCh38
rs1017693104 Chr.1: 23778845-23778845 on Build GRCh38
rs934488782 Chr.1: 23778843-23778843 on Build GRCh38
rs1027928214 Chr.1: 23778800-23778800 on Build GRCh38
rs560039338 Chr.1: 23778753-23778753 on Build GRCh38
rs924504083 Chr.1: 23778838-23778838 on Build GRCh38
rs546910390 Chr.1: 23778808-23778808 on Build GRCh38
rs747967493 Chr.1: 23778734-23778756 on Build GRCh38
rs200606364 Chr.1: 23778739-23778739 on Build GRCh38
rs373015089 Chr.1: 23778656-23778656 on Build GRCh38
rs993397308 Chr.1: 23778778-23778778 on Build GRCh38
rs986079638 Chr.1: 23778754-23778754 on Build GRCh38
rs115234608 Chr.1: 23778805-23778805 on Build GRCh38
rs753285711 Chr.1: 23778709-23778709 on Build GRCh38
rs1007685613 Chr.1: 23778832-23778832 on Build GRCh38
rs764096139 Chr.1: 23778701-23778701 on Build GRCh38
rs939029440 Chr.1: 23778671-23778671 on Build GRCh38
rs764795180 Chr.1: 23778741-23778741 on Build GRCh38
rs966083363 Chr.1: 23778877-23778877 on Build GRCh38
rs775378415 Chr.1: 23778655-23778655 on Build GRCh38

View Full Product Details