Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 105854877-105855136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCK2
Entrez Gene ID
8440
Gene Name
NCK adaptor protein 2
Gene Aliases
GRB4, NCKbeta
UniGene
Hs.529244
Gene Chromosome Location
Chr.2: 105744649-105894274 on Build GRCh38
Transcripts
NM_001004720.2 NM_001004722.3 NM_003581.4 XM_006712797.3 XM_011511991.2 XM_011511992.2 XM_017005103.1 XM_017005104.1 XM_017005105.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4705871 c.66C>A p.D22E Chr.2: 105855129-105855129 on Build GRCh38
5811891 c.43G>T p.A15S Chr.2: 105855106-105855106 on Build GRCh38
440935 c.34G>T p.D12Y Chr.2: 105855097-105855097 on Build GRCh38
4602061 c.28A>C p.K10Q Chr.2: 105855091-105855091 on Build GRCh38
6722905 c.43G>A p.A15T Chr.2: 105855106-105855106 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756342728 Chr.2: 105855042-105855042 on Build GRCh38
rs942777372 Chr.2: 105854938-105854938 on Build GRCh38
rs982875773 Chr.2: 105854993-105854993 on Build GRCh38
rs6543345 Chr.2: 105854937-105854937 on Build GRCh38
rs1003548097 Chr.2: 105855058-105855058 on Build GRCh38
rs201418674 Chr.2: 105855080-105855080 on Build GRCh38
rs929884555 Chr.2: 105854927-105854927 on Build GRCh38
rs560584160 Chr.2: 105855007-105855007 on Build GRCh38
rs114905728 Chr.2: 105855006-105855006 on Build GRCh38
rs769339535 Chr.2: 105855082-105855082 on Build GRCh38
rs775518109 Chr.2: 105854935-105854935 on Build GRCh38
rs768027105 Chr.2: 105855106-105855106 on Build GRCh38
rs774773148 Chr.2: 105855092-105855092 on Build GRCh38
rs771902940 Chr.2: 105855020-105855020 on Build GRCh38
rs772957707 Chr.2: 105855034-105855034 on Build GRCh38
rs1039745901 Chr.2: 105854983-105854983 on Build GRCh38
rs762382562 Chr.2: 105855105-105855105 on Build GRCh38
rs917259503 Chr.2: 105854981-105854981 on Build GRCh38
rs533244550 Chr.2: 105854968-105854968 on Build GRCh38
rs750532600 Chr.2: 105855014-105855014 on Build GRCh38
rs758898610 Chr.2: 105855081-105855081 on Build GRCh38
rs918507791 Chr.2: 105854912-105854912 on Build GRCh38
rs1037580549 Chr.2: 105855022-105855022 on Build GRCh38
rs746692898 Chr.2: 105855043-105855043 on Build GRCh38
rs191132885 Chr.2: 105854911-105854911 on Build GRCh38
rs959247075 Chr.2: 105854931-105854931 on Build GRCh38
rs950054781 Chr.2: 105854992-105854992 on Build GRCh38
rs770518975 Chr.2: 105855050-105855050 on Build GRCh38

View Full Product Details