Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141419148-141419373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGA10
Entrez Gene ID
56106
Gene Name
protocadherin gamma subfamily A, 10
Gene Aliases
PCDH-GAMMA-A10
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141413176-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGA6
Entrez Gene ID
56109
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
PCDH-GAMMA-A6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141374084-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGA7
Entrez Gene ID
56108
Gene Name
protocadherin gamma subfamily A, 7
Gene Aliases
PCDH-GAMMA-A7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141382739-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGA8
Entrez Gene ID
9708
Gene Name
protocadherin gamma subfamily A, 8
Gene Aliases
PCDH-GAMMA-A8
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141391916-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGA9
Entrez Gene ID
56107
Gene Name
protocadherin gamma subfamily A, 9
Gene Aliases
PCDH-GAMMA-A9
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141402953-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGB3
Entrez Gene ID
56102
Gene Name
protocadherin gamma subfamily B, 3
Gene Aliases
PCDH-GAMMA-B3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141370264-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGB4
Entrez Gene ID
8641
Gene Name
protocadherin gamma subfamily B, 4
Gene Aliases
CDH20, FIB2, PCDH-GAMMA-B4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141387885-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGB5
Entrez Gene ID
56101
Gene Name
protocadherin gamma subfamily B, 5
Gene Aliases
PCDH-GAMMA-B5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141398128-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGB6
Entrez Gene ID
56100
Gene Name
protocadherin gamma subfamily B, 6
Gene Aliases
PCDH-GAMMA-B6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141408203-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Gene Symbol
PCDHGB7
Entrez Gene ID
56099
Gene Name
protocadherin gamma subfamily B, 7
Gene Aliases
ME6, PCDH-GAMMA-B7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141417647-141512981 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_032101.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5425865 c.1510T>G p.Y504D Chr.5: 141419369-141419369 on Build GRCh38
4465228 c.1372C>T p.L458L Chr.5: 141419231-141419231 on Build GRCh38
482211 c.1443C>G p.F481L Chr.5: 141419302-141419302 on Build GRCh38
1063284 c.1453G>A p.G485S Chr.5: 141419312-141419312 on Build GRCh38
590097 c.1344C>T p.N448N Chr.5: 141419203-141419203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201698529 Chr.5: 141419203-141419203 on Build GRCh38
rs777238100 Chr.5: 141419275-141419275 on Build GRCh38
rs370480327 Chr.5: 141419322-141419322 on Build GRCh38
rs202038869 Chr.5: 141419350-141419350 on Build GRCh38
rs751868316 Chr.5: 141419273-141419273 on Build GRCh38
rs775328616 Chr.5: 141419316-141419316 on Build GRCh38
rs762064534 Chr.5: 141419336-141419336 on Build GRCh38
rs890162404 Chr.5: 141419351-141419351 on Build GRCh38
rs549801775 Chr.5: 141419346-141419346 on Build GRCh38
rs755798236 Chr.5: 141419274-141419274 on Build GRCh38
rs778360128 Chr.5: 141419305-141419305 on Build GRCh38
rs185228661 Chr.5: 141419175-141419175 on Build GRCh38
rs560134083 Chr.5: 141419197-141419197 on Build GRCh38
rs374093302 Chr.5: 141419239-141419239 on Build GRCh38
rs758796140 Chr.5: 141419170-141419170 on Build GRCh38
rs774592605 Chr.5: 141419328-141419328 on Build GRCh38
rs560904796 Chr.5: 141419226-141419226 on Build GRCh38
rs768622826 Chr.5: 141419326-141419326 on Build GRCh38
rs770303087 Chr.5: 141419294-141419294 on Build GRCh38
rs769321564 Chr.5: 141419181-141419181 on Build GRCh38
rs761014077 Chr.5: 141419228-141419228 on Build GRCh38
rs764216382 Chr.5: 141419236-141419236 on Build GRCh38
rs758952830 Chr.5: 141419261-141419261 on Build GRCh38
rs765877993 Chr.5: 141419259-141419259 on Build GRCh38
rs772847766 Chr.5: 141419192-141419192 on Build GRCh38
rs748856660 Chr.5: 141419293-141419293 on Build GRCh38
rs771892411 Chr.5: 141419215-141419215 on Build GRCh38
rs749331717 Chr.5: 141419247-141419259 on Build GRCh38
rs928013251 Chr.5: 141419235-141419235 on Build GRCh38
rs761740232 Chr.5: 141419258-141419258 on Build GRCh38
rs35892780 Chr.5: 141419312-141419312 on Build GRCh38
rs776677714 Chr.5: 141419329-141419329 on Build GRCh38
rs371077803 Chr.5: 141419180-141419180 on Build GRCh38
rs111954647 Chr.5: 141419345-141419345 on Build GRCh38
rs959206942 Chr.5: 141419243-141419243 on Build GRCh38
rs541061354 Chr.5: 141419209-141419209 on Build GRCh38
rs753956971 Chr.5: 141419240-141419240 on Build GRCh38
rs745852942 Chr.5: 141419311-141419311 on Build GRCh38

View Full Product Details