Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41838798-41839051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYPD4
Entrez Gene ID
147719
Gene Name
LY6/PLAUR domain containing 4
Gene Aliases
SMR
UniGene
Hs.382361
Gene Chromosome Location
Chr.19: 41833686-41844771 on Build GRCh38
Transcripts
NM_001291419.1 NM_173506.6 XM_005258543.3 XM_005278383.4 XM_005278385.4 XM_005278387.4 XM_005278388.3 XM_006723035.2 XM_011526483.2 XM_011526484.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5921467 c.69G>A p.R23R Chr.19: 41839023-41839023 on Build GRCh38
3892678 c.201C>T p.F67F Chr.19: 41838891-41838891 on Build GRCh38
1612305 c.113G>T p.R38I Chr.19: 41838979-41838979 on Build GRCh38
4078576 c.92A>G p.E31G Chr.19: 41839000-41839000 on Build GRCh38
5525208 c.169C>T p.L57L Chr.19: 41838923-41838923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782687678 Chr.19: 41839017-41839017 on Build GRCh38
rs782088156 Chr.19: 41838932-41838932 on Build GRCh38
rs368052508 Chr.19: 41838831-41838831 on Build GRCh38
rs782600420 Chr.19: 41838986-41838986 on Build GRCh38
rs782169686 Chr.19: 41838841-41838841 on Build GRCh38
rs150537336 Chr.19: 41838935-41838935 on Build GRCh38
rs202011969 Chr.19: 41838976-41838976 on Build GRCh38
rs781913877 Chr.19: 41839022-41839022 on Build GRCh38
rs782353361 Chr.19: 41839021-41839021 on Build GRCh38
rs782343746 Chr.19: 41838896-41838896 on Build GRCh38
rs782139533 Chr.19: 41838925-41838925 on Build GRCh38
rs939139055 Chr.19: 41838884-41838884 on Build GRCh38
rs368266342 Chr.19: 41838979-41838979 on Build GRCh38
rs782820684 Chr.19: 41839025-41839025 on Build GRCh38
rs139542363 Chr.19: 41839010-41839010 on Build GRCh38
rs371331001 Chr.19: 41838853-41838853 on Build GRCh38
rs781897511 Chr.19: 41838960-41838960 on Build GRCh38
rs374515505 Chr.19: 41838857-41838857 on Build GRCh38
rs112970900 Chr.19: 41838878-41838878 on Build GRCh38
rs371966671 Chr.19: 41838892-41838892 on Build GRCh38
rs781940438 Chr.19: 41838900-41838900 on Build GRCh38
rs782158115 Chr.19: 41839023-41839023 on Build GRCh38
rs782269844 Chr.19: 41838881-41838881 on Build GRCh38
rs542631666 Chr.19: 41838872-41838872 on Build GRCh38
rs368030545 Chr.19: 41838880-41838880 on Build GRCh38
rs138135884 Chr.19: 41838956-41838956 on Build GRCh38
rs1044339869 Chr.19: 41838914-41838914 on Build GRCh38
rs150227643 Chr.19: 41838885-41838885 on Build GRCh38
rs371971494 Chr.19: 41838990-41838990 on Build GRCh38
rs782656233 Chr.19: 41839002-41839002 on Build GRCh38
rs782048116 Chr.19: 41838901-41838901 on Build GRCh38
rs527657318 Chr.19: 41838994-41838994 on Build GRCh38
rs782523801 Chr.19: 41838964-41838964 on Build GRCh38
rs113358585 Chr.19: 41838879-41838879 on Build GRCh38
rs200441266 Chr.19: 41838913-41838913 on Build GRCh38
rs1040605661 Chr.19: 41838927-41838927 on Build GRCh38
rs112654847 Chr.19: 41838919-41838919 on Build GRCh38
rs562198016 Chr.19: 41838895-41838895 on Build GRCh38
rs781934468 Chr.19: 41838930-41838931 on Build GRCh38
rs1054811504 Chr.19: 41838886-41838886 on Build GRCh38
rs781997244 Chr.19: 41839008-41839008 on Build GRCh38
rs78978026 Chr.19: 41838930-41838930 on Build GRCh38
rs1000084791 Chr.19: 41838926-41838926 on Build GRCh38
rs927698236 Chr.19: 41838852-41838852 on Build GRCh38
rs782787878 Chr.19: 41838965-41838965 on Build GRCh38
rs782486302 Chr.19: 41838981-41838981 on Build GRCh38
rs782092299 Chr.19: 41838944-41838944 on Build GRCh38
rs782311545 Chr.19: 41838868-41838869 on Build GRCh38
rs984405021 Chr.19: 41838823-41838823 on Build GRCh38
rs549358411 Chr.19: 41839003-41839003 on Build GRCh38
rs782800342 Chr.19: 41838842-41838842 on Build GRCh38
rs201334161 Chr.19: 41838850-41838850 on Build GRCh38
rs782366023 Chr.19: 41839004-41839004 on Build GRCh38
rs10414356 Chr.19: 41838849-41838849 on Build GRCh38

View Full Product Details