Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68403446-68403679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRP5
Entrez Gene ID
4041
Gene Name
LDL receptor related protein 5
Gene Aliases
BMND1, EVR1, EVR4, HBM, LR3, LRP-5, LRP7, OPPG, OPS, OPTA1, VBCH2
UniGene
Hs.6347
Gene Chromosome Location
Chr.11: 68298866-68449275 on Build GRCh38
Transcripts
NM_001291902.1 NM_002335.3 XM_005273994.2 XM_011545029.1 XM_011545030.1 XM_011545031.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7262510 c.1693C>T p.R565C Chr.11: 68403591-68403591 on Build GRCh38
4530525 c.1705G>A p.E569K Chr.11: 68403603-68403603 on Build GRCh38
1676248 c.1689G>A p.W563* Chr.11: 68403587-68403587 on Build GRCh38
213068 c.1652T>G p.F551C Chr.11: 68403550-68403550 on Build GRCh38
4403192 c.1645T>A p.F549I Chr.11: 68403543-68403543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200627198 Chr.11: 68403602-68403602 on Build GRCh38
rs972629478 Chr.11: 68403515-68403515 on Build GRCh38
rs80358311 Chr.11: 68403546-68403546 on Build GRCh38
rs765468063 Chr.11: 68403630-68403630 on Build GRCh38
rs545382 Chr.11: 68403545-68403545 on Build GRCh38
rs759331617 Chr.11: 68403513-68403513 on Build GRCh38
rs397514663 Chr.11: 68403553-68403553 on Build GRCh38
rs776728388 Chr.11: 68403619-68403619 on Build GRCh38
rs566952681 Chr.11: 68403525-68403525 on Build GRCh38
rs146285115 Chr.11: 68403554-68403554 on Build GRCh38
rs770292689 Chr.11: 68403510-68403510 on Build GRCh38
rs184457951 Chr.11: 68403560-68403560 on Build GRCh38
rs866907018 Chr.11: 68403550-68403550 on Build GRCh38
rs767464846 Chr.11: 68403611-68403611 on Build GRCh38
rs139408531 Chr.11: 68403581-68403581 on Build GRCh38
rs758798637 Chr.11: 68403635-68403635 on Build GRCh38
rs761424454 Chr.11: 68403527-68403527 on Build GRCh38
rs774083040 Chr.11: 68403536-68403536 on Build GRCh38
rs999670238 Chr.11: 68403639-68403639 on Build GRCh38
rs138692892 Chr.11: 68403511-68403511 on Build GRCh38
rs911000449 Chr.11: 68403592-68403592 on Build GRCh38
rs746554984 Chr.11: 68403505-68403505 on Build GRCh38
rs199707305 Chr.11: 68403605-68403605 on Build GRCh38
rs149524398 Chr.11: 68403636-68403636 on Build GRCh38
rs762794790 Chr.11: 68403531-68403531 on Build GRCh38
rs775233316 Chr.11: 68403594-68403594 on Build GRCh38
rs764112300 Chr.11: 68403533-68403533 on Build GRCh38
rs751940504 Chr.11: 68403656-68403656 on Build GRCh38
rs201576246 Chr.11: 68403540-68403540 on Build GRCh38
rs80358310 Chr.11: 68403502-68403502 on Build GRCh38
rs587783024 Chr.11: 68403595-68403595 on Build GRCh38
rs1056743758 Chr.11: 68403642-68403642 on Build GRCh38
rs574974332 Chr.11: 68403622-68403622 on Build GRCh38
rs896721075 Chr.11: 68403648-68403648 on Build GRCh38
rs141896162 Chr.11: 68403503-68403503 on Build GRCh38
rs757754971 Chr.11: 68403659-68403659 on Build GRCh38
rs150209973 Chr.11: 68403506-68403506 on Build GRCh38
rs747408714 Chr.11: 68403564-68403564 on Build GRCh38
rs769557748 Chr.11: 68403522-68403522 on Build GRCh38
rs377144001 Chr.11: 68403578-68403578 on Build GRCh38
rs757777983 Chr.11: 68403575-68403575 on Build GRCh38
rs202067798 Chr.11: 68403474-68403474 on Build GRCh38
rs779303324 Chr.11: 68403481-68403481 on Build GRCh38
rs200827533 Chr.11: 68403473-68403473 on Build GRCh38
rs745350421 Chr.11: 68403591-68403591 on Build GRCh38
rs768268948 Chr.11: 68403603-68403603 on Build GRCh38
rs752862568 Chr.11: 68403631-68403631 on Build GRCh38
rs748487092 Chr.11: 68403491-68403491 on Build GRCh38
rs774097621 Chr.11: 68403534-68403534 on Build GRCh38
rs778361337 Chr.11: 68403563-68403563 on Build GRCh38
rs80358312 Chr.11: 68403607-68403607 on Build GRCh38
rs121908665 Chr.11: 68403606-68403606 on Build GRCh38

View Full Product Details