Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67353202-67353475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100130987
Entrez Gene ID
100130987
Gene Name
uncharacterized LOC100130987
Gene Aliases
-
UniGene
Hs.664585
Gene Chromosome Location
Chr.11: 67317839-67391687 on Build GRCh38
Transcripts
NR_024469.1 NM_001256870.1 NM_021173.4 NR_046411.1 NR_046412.1 NR_046413.1

Gene Symbol
POLD4
Entrez Gene ID
57804
Gene Name
DNA polymerase delta 4, accessory subunit
Gene Aliases
POLDS, p12
UniGene
Hs.523829
Gene Chromosome Location
Chr.11: 67350765-67353596 on Build GRCh38
Transcripts
NR_024469.1 NM_001256870.1 NM_021173.4 NR_046411.1 NR_046412.1 NR_046413.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5729784 c.88C>T p.P30S Chr.11: 67353312-67353312 on Build GRCh38
7313410 c.86C>A p.A29E Chr.11: 67353314-67353314 on Build GRCh38
3869981 c.18C>T p.L6L Chr.11: 67353382-67353382 on Build GRCh38
5477766 c.83T>G p.L28R Chr.11: 67353317-67353317 on Build GRCh38
3764338 c.2T>G p.M1R Chr.11: 67353398-67353398 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs932377370 Chr.11: 67353453-67353453 on Build GRCh38
rs778149868 Chr.11: 67353365-67353365 on Build GRCh38
rs769491961 Chr.11: 67353420-67353420 on Build GRCh38
rs944001533 Chr.11: 67353364-67353364 on Build GRCh38
rs1014013415 Chr.11: 67353253-67353253 on Build GRCh38
rs771361847 Chr.11: 67353371-67353371 on Build GRCh38
rs754485355 Chr.11: 67353319-67353319 on Build GRCh38
rs911689647 Chr.11: 67353278-67353278 on Build GRCh38
rs747903396 Chr.11: 67353347-67353347 on Build GRCh38
rs748439555 Chr.11: 67353421-67353423 on Build GRCh38
rs768518371 Chr.11: 67353254-67353254 on Build GRCh38
rs748993149 Chr.11: 67353427-67353427 on Build GRCh38
rs747518956 Chr.11: 67353278-67353278 on Build GRCh38
rs778829795 Chr.11: 67353266-67353266 on Build GRCh38
rs772492719 Chr.11: 67353274-67353274 on Build GRCh38
rs531618182 Chr.11: 67353340-67353340 on Build GRCh38
rs1042844120 Chr.11: 67353457-67353457 on Build GRCh38
rs1031278504 Chr.11: 67353221-67353221 on Build GRCh38
rs560690127 Chr.11: 67353229-67353229 on Build GRCh38
rs775264611 Chr.11: 67353423-67353423 on Build GRCh38
rs935669164 Chr.11: 67353395-67353395 on Build GRCh38
rs779594503 Chr.11: 67353273-67353275 on Build GRCh38
rs745371682 Chr.11: 67353410-67353410 on Build GRCh38
rs780693051 Chr.11: 67353375-67353375 on Build GRCh38
rs776034868 Chr.11: 67353293-67353293 on Build GRCh38
rs199992452 Chr.11: 67353328-67353328 on Build GRCh38
rs55855949 Chr.11: 67353307-67353307 on Build GRCh38
rs770442598 Chr.11: 67353286-67353286 on Build GRCh38
rs769580420 Chr.11: 67353310-67353310 on Build GRCh38
rs758740392 Chr.11: 67353350-67353350 on Build GRCh38
rs748518077 Chr.11: 67353259-67353259 on Build GRCh38
rs766436446 Chr.11: 67353338-67353338 on Build GRCh38
rs778239887 Chr.11: 67353275-67353275 on Build GRCh38
rs775354650 Chr.11: 67353316-67353316 on Build GRCh38
rs920960565 Chr.11: 67353386-67353386 on Build GRCh38
rs980903205 Chr.11: 67353309-67353309 on Build GRCh38
rs753879282 Chr.11: 67353339-67353339 on Build GRCh38
rs779104220 Chr.11: 67353342-67353342 on Build GRCh38
rs371109758 Chr.11: 67353305-67353305 on Build GRCh38
rs150889244 Chr.11: 67353324-67353324 on Build GRCh38
rs779192794 Chr.11: 67353252-67353252 on Build GRCh38
rs369600661 Chr.11: 67353227-67353227 on Build GRCh38
rs200868910 Chr.11: 67353321-67353321 on Build GRCh38
rs908473394 Chr.11: 67353334-67353334 on Build GRCh38
rs753489289 Chr.11: 67353310-67353310 on Build GRCh38

View Full Product Details