Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102138952-102139187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPRC1
Entrez Gene ID
23082
Gene Name
peroxisome proliferator-activated receptor gamma, coactivator-related 1
Gene Aliases
PRC
UniGene
Hs.533551
Gene Chromosome Location
Chr.10: 102132994-102150333 on Build GRCh38
Transcripts
NM_001288727.1 NM_001288728.1 NM_015062.4 XM_005269656.3 XM_006717731.3 XM_011539549.2 XM_011539551.2 XM_011539552.2 XM_017015982.1 XM_017015983.1 XM_017015984.1 XM_017015985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4504548 c.668C>T p.P223L Chr.10: 102139176-102139176 on Build GRCh38
3665454 c.671G>A p.S224N Chr.10: 102139179-102139179 on Build GRCh38
5421657 c.615C>T p.P205P Chr.10: 102139123-102139123 on Build GRCh38
6488428 c.592G>T p.V198F Chr.10: 102139100-102139100 on Build GRCh38
5926744 c.614C>T p.P205L Chr.10: 102139122-102139122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760518646 Chr.10: 102139054-102139054 on Build GRCh38
rs762621105 Chr.10: 102139019-102139021 on Build GRCh38
rs575761739 Chr.10: 102138985-102138985 on Build GRCh38
rs17847382 Chr.10: 102139079-102139079 on Build GRCh38
rs778506664 Chr.10: 102139014-102139014 on Build GRCh38
rs757684751 Chr.10: 102138975-102138975 on Build GRCh38
rs17847383 Chr.10: 102139132-102139132 on Build GRCh38
rs756674657 Chr.10: 102139121-102139121 on Build GRCh38
rs373478752 Chr.10: 102138973-102138973 on Build GRCh38
rs751829107 Chr.10: 102139094-102139094 on Build GRCh38
rs776219118 Chr.10: 102139076-102139076 on Build GRCh38
rs770429418 Chr.10: 102139155-102139155 on Build GRCh38
rs948526617 Chr.10: 102139055-102139055 on Build GRCh38
rs1053572547 Chr.10: 102139033-102139033 on Build GRCh38
rs747616605 Chr.10: 102139020-102139020 on Build GRCh38
rs746664279 Chr.10: 102139146-102139146 on Build GRCh38
rs752869049 Chr.10: 102139091-102139091 on Build GRCh38
rs757888892 Chr.10: 102139143-102139143 on Build GRCh38
rs763649697 Chr.10: 102139069-102139071 on Build GRCh38
rs750705554 Chr.10: 102139115-102139115 on Build GRCh38
rs373045453 Chr.10: 102139123-102139123 on Build GRCh38
rs770753120 Chr.10: 102139064-102139064 on Build GRCh38
rs765048524 Chr.10: 102139086-102139086 on Build GRCh38
rs199712231 Chr.10: 102139144-102139144 on Build GRCh38
rs929027213 Chr.10: 102139125-102139125 on Build GRCh38
rs771749685 Chr.10: 102139025-102139025 on Build GRCh38
rs866875893 Chr.10: 102139154-102139154 on Build GRCh38
rs757490022 Chr.10: 102139108-102139108 on Build GRCh38
rs990022320 Chr.10: 102139013-102139013 on Build GRCh38
rs781617641 Chr.10: 102138980-102138980 on Build GRCh38
rs764472672 Chr.10: 102139093-102139093 on Build GRCh38
rs1021130777 Chr.10: 102139122-102139122 on Build GRCh38
rs772827671 Chr.10: 102139030-102139030 on Build GRCh38
rs377421036 Chr.10: 102138991-102138991 on Build GRCh38

View Full Product Details