Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22120279-22120542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HR
Entrez Gene ID
55806
Gene Name
hair growth associated
Gene Aliases
ALUNC, AU, HSA277165, HYPT4, MUHH, MUHH1
UniGene
Hs.272367
Gene Chromosome Location
Chr.8: 22114419-22131053 on Build GRCh38
Transcripts
NM_005144.4 NM_018411.4 XM_005273569.1 XM_006716367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5710918 c.2717C>T p.P906L Chr.8: 22120401-22120401 on Build GRCh38
376456 c.2678C>T p.A893V Chr.8: 22120440-22120440 on Build GRCh38
6112791 c.2699C>T p.A900V Chr.8: 22120419-22120419 on Build GRCh38
3647211 c.2765C>T p.S922F Chr.8: 22120353-22120353 on Build GRCh38
4483577 c.2712C>T p.L904L Chr.8: 22120406-22120406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11990421 Chr.8: 22120347-22120347 on Build GRCh38
rs560349127 Chr.8: 22120296-22120296 on Build GRCh38
rs367558699 Chr.8: 22120420-22120420 on Build GRCh38
rs11990450 Chr.8: 22120384-22120384 on Build GRCh38
rs998329974 Chr.8: 22120454-22120454 on Build GRCh38
rs11990451 Chr.8: 22120385-22120385 on Build GRCh38
rs768618620 Chr.8: 22120421-22120421 on Build GRCh38
rs143947629 Chr.8: 22120455-22120455 on Build GRCh38
rs755592901 Chr.8: 22120382-22120382 on Build GRCh38
rs773764015 Chr.8: 22120341-22120341 on Build GRCh38
rs745691683 Chr.8: 22120405-22120405 on Build GRCh38
rs1005268498 Chr.8: 22120406-22120406 on Build GRCh38
rs1015333064 Chr.8: 22120410-22120410 on Build GRCh38
rs780149768 Chr.8: 22120418-22120418 on Build GRCh38
rs776833962 Chr.8: 22120434-22120434 on Build GRCh38
rs375878986 Chr.8: 22120342-22120342 on Build GRCh38
rs945747109 Chr.8: 22120393-22120393 on Build GRCh38
rs778646464 Chr.8: 22120400-22120400 on Build GRCh38
rs895762780 Chr.8: 22120331-22120331 on Build GRCh38
rs771886990 Chr.8: 22120482-22120482 on Build GRCh38
rs376800273 Chr.8: 22120515-22120515 on Build GRCh38
rs765178685 Chr.8: 22120502-22120502 on Build GRCh38
rs777004940 Chr.8: 22120320-22120320 on Build GRCh38
rs372128408 Chr.8: 22120461-22120461 on Build GRCh38
rs116376389 Chr.8: 22120501-22120501 on Build GRCh38
rs779962253 Chr.8: 22120522-22120522 on Build GRCh38
rs758563839 Chr.8: 22120408-22120408 on Build GRCh38
rs117242897 Chr.8: 22120511-22120511 on Build GRCh38
rs767109447 Chr.8: 22120349-22120349 on Build GRCh38
rs760433405 Chr.8: 22120463-22120463 on Build GRCh38
rs552218948 Chr.8: 22120358-22120358 on Build GRCh38
rs758368200 Chr.8: 22120339-22120340 on Build GRCh38
rs773674017 Chr.8: 22120457-22120457 on Build GRCh38
rs185501052 Chr.8: 22120419-22120419 on Build GRCh38
rs888156911 Chr.8: 22120398-22120398 on Build GRCh38
rs868314387 Chr.8: 22120422-22120422 on Build GRCh38
rs748645922 Chr.8: 22120440-22120440 on Build GRCh38
rs569125827 Chr.8: 22120496-22120496 on Build GRCh38
rs753705092 Chr.8: 22120392-22120392 on Build GRCh38
rs757232733 Chr.8: 22120394-22120394 on Build GRCh38
rs765686683 Chr.8: 22120322-22120322 on Build GRCh38
rs750157785 Chr.8: 22120513-22120513 on Build GRCh38

View Full Product Details