Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 23954592-23954837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR1D2
Entrez Gene ID
9975
Gene Name
nuclear receptor subfamily 1 group D member 2
Gene Aliases
BD73, EAR-1R, RVR
UniGene
Hs.37288
Gene Chromosome Location
Chr.3: 23945260-23980618 on Build GRCh38
Transcripts
NM_001145425.1 NM_005126.4 NR_110524.1 XM_006713451.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6729041 c.111C>T p.S37S Chr.3: 23954631-23954631 on Build GRCh38
1745092 c.207delT p.N69fs*4 Chr.3: 23954727-23954727 on Build GRCh38
6729042 c.168G>T p.K56N Chr.3: 23954688-23954688 on Build GRCh38
6047075 c.174T>C p.G58G Chr.3: 23954694-23954694 on Build GRCh38
1259418 c.181G>A p.A61T Chr.3: 23954701-23954701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036545367 Chr.3: 23954714-23954714 on Build GRCh38
rs1022465161 Chr.3: 23954800-23954800 on Build GRCh38
rs762861304 Chr.3: 23954750-23954750 on Build GRCh38
rs931994383 Chr.3: 23954644-23954644 on Build GRCh38
rs767076379 Chr.3: 23954770-23954770 on Build GRCh38
rs769593623 Chr.3: 23954657-23954657 on Build GRCh38
rs528383367 Chr.3: 23954653-23954653 on Build GRCh38
rs371828572 Chr.3: 23954628-23954628 on Build GRCh38
rs976099541 Chr.3: 23954619-23954619 on Build GRCh38
rs764920839 Chr.3: 23954731-23954731 on Build GRCh38
rs780084653 Chr.3: 23954739-23954739 on Build GRCh38
rs141523313 Chr.3: 23954733-23954733 on Build GRCh38
rs762916242 Chr.3: 23954672-23954672 on Build GRCh38
rs778365984 Chr.3: 23954625-23954625 on Build GRCh38
rs746002238 Chr.3: 23954732-23954732 on Build GRCh38
rs767011386 Chr.3: 23954696-23954696 on Build GRCh38
rs757889206 Chr.3: 23954715-23954715 on Build GRCh38
rs764534052 Chr.3: 23954811-23954811 on Build GRCh38
rs747281208 Chr.3: 23954740-23954740 on Build GRCh38
rs906489074 Chr.3: 23954716-23954716 on Build GRCh38
rs770917860 Chr.3: 23954754-23954754 on Build GRCh38
rs201515529 Chr.3: 23954698-23954698 on Build GRCh38
rs568010643 Chr.3: 23954705-23954705 on Build GRCh38
rs923283545 Chr.3: 23954627-23954627 on Build GRCh38
rs769541377 Chr.3: 23954744-23954744 on Build GRCh38
rs751084374 Chr.3: 23954687-23954687 on Build GRCh38
rs563905774 Chr.3: 23954617-23954617 on Build GRCh38
rs143811972 Chr.3: 23954693-23954693 on Build GRCh38
rs760442210 Chr.3: 23954806-23954806 on Build GRCh38
rs746082481 Chr.3: 23954631-23954631 on Build GRCh38
rs772242470 Chr.3: 23954643-23954643 on Build GRCh38
rs773037418 Chr.3: 23954746-23954746 on Build GRCh38
rs752250438 Chr.3: 23954771-23954771 on Build GRCh38
rs757278641 Chr.3: 23954713-23954713 on Build GRCh38
rs775035300 Chr.3: 23954623-23954625 on Build GRCh38
rs956649146 Chr.3: 23954753-23954753 on Build GRCh38
rs776481412 Chr.3: 23954651-23954651 on Build GRCh38
rs143411191 Chr.3: 23954700-23954700 on Build GRCh38
rs758912664 Chr.3: 23954762-23954762 on Build GRCh38
rs765849666 Chr.3: 23954675-23954675 on Build GRCh38
rs754367426 Chr.3: 23954706-23954706 on Build GRCh38
rs773086538 Chr.3: 23954669-23954669 on Build GRCh38
rs774345944 Chr.3: 23954757-23954757 on Build GRCh38
rs986826852 Chr.3: 23954683-23954683 on Build GRCh38

View Full Product Details