Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57629641-57629914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCOM1
Entrez Gene ID
145781
Gene Name
GRINL1A complex locus 1
Gene Aliases
GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57717557 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Gene Symbol
MYZAP
Entrez Gene ID
100820829
Gene Name
myocardial zonula adherens protein
Gene Aliases
GCOM1, Gup, MYOZAP
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57685364 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1493483 c.557G>A p.S186N Chr.15: 57629733-57629733 on Build GRCh38
5955540 c.494A>T p.Q165L Chr.15: 57629763-57629763 on Build GRCh38
5585871 c.622G>A p.E208K Chr.15: 57629798-57629798 on Build GRCh38
1301255 c.566A>T p.K189M Chr.15: 57629742-57629742 on Build GRCh38
5585872 c.529G>A p.E177K Chr.15: 57629798-57629798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755738804 Chr.15: 57629690-57629690 on Build GRCh38
rs148618731 Chr.15: 57629714-57629714 on Build GRCh38
rs113448249 Chr.15: 57629776-57629776 on Build GRCh38
rs966174304 Chr.15: 57629665-57629665 on Build GRCh38
rs768033320 Chr.15: 57629878-57629878 on Build GRCh38
rs368082220 Chr.15: 57629763-57629763 on Build GRCh38
rs767790356 Chr.15: 57629773-57629773 on Build GRCh38
rs746777758 Chr.15: 57629818-57629818 on Build GRCh38
rs774504358 Chr.15: 57629695-57629695 on Build GRCh38
rs764542251 Chr.15: 57629885-57629885 on Build GRCh38
rs374583807 Chr.15: 57629698-57629698 on Build GRCh38
rs572650003 Chr.15: 57629752-57629752 on Build GRCh38
rs769353202 Chr.15: 57629776-57629786 on Build GRCh38
rs753520646 Chr.15: 57629790-57629790 on Build GRCh38
rs144837547 Chr.15: 57629662-57629662 on Build GRCh38
rs550096099 Chr.15: 57629730-57629730 on Build GRCh38
rs114058997 Chr.15: 57629667-57629667 on Build GRCh38
rs745798322 Chr.15: 57629812-57629812 on Build GRCh38
rs200330646 Chr.15: 57629769-57629769 on Build GRCh38
rs371814833 Chr.15: 57629770-57629770 on Build GRCh38
rs755755953 Chr.15: 57629781-57629781 on Build GRCh38
rs756886268 Chr.15: 57629791-57629791 on Build GRCh38
rs935274009 Chr.15: 57629798-57629798 on Build GRCh38
rs778562303 Chr.15: 57629792-57629792 on Build GRCh38
rs755344818 Chr.15: 57629668-57629668 on Build GRCh38
rs1053521813 Chr.15: 57629802-57629802 on Build GRCh38
rs80174915 Chr.15: 57629817-57629817 on Build GRCh38
rs200077771 Chr.15: 57629820-57629820 on Build GRCh38
rs771060878 Chr.15: 57629851-57629851 on Build GRCh38
rs761954649 Chr.15: 57629720-57629720 on Build GRCh38
rs375537951 Chr.15: 57629784-57629784 on Build GRCh38
rs747505374 Chr.15: 57629666-57629666 on Build GRCh38
rs770536891 Chr.15: 57629707-57629707 on Build GRCh38
rs759986886 Chr.15: 57629868-57629868 on Build GRCh38
rs541541184 Chr.15: 57629855-57629855 on Build GRCh38
rs747818729 Chr.15: 57629686-57629686 on Build GRCh38
rs779044670 Chr.15: 57629710-57629710 on Build GRCh38
rs577998764 Chr.15: 57629857-57629857 on Build GRCh38
rs146699810 Chr.15: 57629879-57629879 on Build GRCh38
rs781604146 Chr.15: 57629675-57629675 on Build GRCh38
rs773469134 Chr.15: 57629823-57629823 on Build GRCh38
rs544553369 Chr.15: 57629815-57629815 on Build GRCh38
rs905996288 Chr.15: 57629876-57629876 on Build GRCh38
rs761174203 Chr.15: 57629884-57629884 on Build GRCh38
rs142052894 Chr.15: 57629824-57629824 on Build GRCh38
rs745806929 Chr.15: 57629711-57629711 on Build GRCh38
rs368405673 Chr.15: 57629778-57629778 on Build GRCh38
rs7171850 Chr.15: 57629661-57629661 on Build GRCh38
rs768406825 Chr.15: 57629822-57629822 on Build GRCh38
rs773311433 Chr.15: 57629744-57629744 on Build GRCh38

View Full Product Details