Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155208921-155209170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTX1
Entrez Gene ID
4580
Gene Name
metaxin 1
Gene Aliases
MTX, MTXN
UniGene
Hs.490874
Gene Chromosome Location
Chr.1: 155208699-155213839 on Build GRCh38
Transcripts
NM_002455.4 NM_198883.3 XM_011509931.1 XM_011509933.1 XM_011509935.1

Gene Symbol
THBS3
Entrez Gene ID
7059
Gene Name
thrombospondin 3
Gene Aliases
TSP3
UniGene
Hs.169875
Gene Chromosome Location
Chr.1: 155195588-155209180 on Build GRCh38
Transcripts
NM_002455.4 NM_198883.3 XM_011509931.1 XM_011509933.1 XM_011509935.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6201732 c.140C>T p.A47V Chr.1: 155208944-155208944 on Build GRCh38
4543070 c.330G>A p.P110P Chr.1: 155209134-155209134 on Build GRCh38
6437139 c.287G>A p.R96H Chr.1: 155209091-155209091 on Build GRCh38
896809 c.126C>A p.R42R Chr.1: 155208930-155208930 on Build GRCh38
5581708 c.282C>T p.V94V Chr.1: 155209086-155209086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779123272 Chr.1: 155209020-155209020 on Build GRCh38
rs964660155 Chr.1: 155209068-155209068 on Build GRCh38
rs201584620 Chr.1: 155208954-155208954 on Build GRCh38
rs757592720 Chr.1: 155209018-155209018 on Build GRCh38
rs760077 Chr.1: 155208991-155208991 on Build GRCh38
rs761274486 Chr.1: 155208982-155208984 on Build GRCh38
rs375558224 Chr.1: 155209005-155209005 on Build GRCh38
rs776070150 Chr.1: 155208977-155208994 on Build GRCh38
rs772698213 Chr.1: 155208940-155208940 on Build GRCh38
rs978443293 Chr.1: 155209087-155209087 on Build GRCh38
rs764988599 Chr.1: 155208978-155208978 on Build GRCh38
rs369692343 Chr.1: 155209022-155209022 on Build GRCh38
rs746453727 Chr.1: 155209132-155209132 on Build GRCh38
rs34921532 Chr.1: 155208955-155208955 on Build GRCh38
rs761436076 Chr.1: 155209153-155209153 on Build GRCh38
rs761559280 Chr.1: 155208970-155208970 on Build GRCh38
rs995818317 Chr.1: 155209073-155209073 on Build GRCh38
rs749607249 Chr.1: 155208941-155208941 on Build GRCh38
rs372914521 Chr.1: 155209100-155209100 on Build GRCh38
rs752653129 Chr.1: 155208996-155208996 on Build GRCh38
rs925575119 Chr.1: 155209097-155209097 on Build GRCh38
rs774846663 Chr.1: 155208956-155208956 on Build GRCh38
rs778092146 Chr.1: 155209009-155209009 on Build GRCh38
rs759927568 Chr.1: 155208950-155208950 on Build GRCh38
rs776162502 Chr.1: 155209152-155209152 on Build GRCh38
rs963396491 Chr.1: 155209055-155209055 on Build GRCh38
rs753515109 Chr.1: 155208966-155208966 on Build GRCh38
rs987304592 Chr.1: 155209137-155209137 on Build GRCh38
rs760296290 Chr.1: 155208962-155208962 on Build GRCh38
rs988942034 Chr.1: 155209113-155209113 on Build GRCh38
rs776890789 Chr.1: 155209032-155209032 on Build GRCh38
rs1044567097 Chr.1: 155209149-155209149 on Build GRCh38
rs774957655 Chr.1: 155208947-155208947 on Build GRCh38
rs749552182 Chr.1: 155209016-155209016 on Build GRCh38
rs758257421 Chr.1: 155208988-155208988 on Build GRCh38
rs769014868 Chr.1: 155208984-155208984 on Build GRCh38
rs763664713 Chr.1: 155208965-155208965 on Build GRCh38
rs1005583605 Chr.1: 155209024-155209024 on Build GRCh38
rs917011173 Chr.1: 155209118-155209118 on Build GRCh38
rs954498050 Chr.1: 155209011-155209011 on Build GRCh38
rs1016501260 Chr.1: 155209049-155209049 on Build GRCh38
rs772450682 Chr.1: 155209046-155209046 on Build GRCh38

View Full Product Details