Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49174250-49174509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLP2
Entrez Gene ID
5355
Gene Name
proteolipid protein 2
Gene Aliases
A4, A4LSB
UniGene
Hs.77422
Gene Chromosome Location
Chr.X: 49171837-49175120 on Build GRCh38
Transcripts
NM_002668.2

SNP Detail

SNP ID Chromosome Location
rs782278583 Chr.X: 49174373-49174373 on Build GRCh38
rs370551031 Chr.X: 49174286-49174286 on Build GRCh38
rs368089500 Chr.X: 49174414-49174414 on Build GRCh38
rs368640397 Chr.X: 49174389-49174389 on Build GRCh38
rs782704183 Chr.X: 49174469-49174469 on Build GRCh38
rs782135920 Chr.X: 49174313-49174313 on Build GRCh38
rs782266678 Chr.X: 49174407-49174407 on Build GRCh38
rs201497020 Chr.X: 49174350-49174350 on Build GRCh38
rs782016426 Chr.X: 49174463-49174463 on Build GRCh38
rs782818987 Chr.X: 49174457-49174457 on Build GRCh38
rs782001907 Chr.X: 49174430-49174430 on Build GRCh38
rs782227811 Chr.X: 49174419-49174419 on Build GRCh38
rs138735148 Chr.X: 49174423-49174423 on Build GRCh38
rs55794588 Chr.X: 49174294-49174294 on Build GRCh38
rs782109629 Chr.X: 49174453-49174453 on Build GRCh38
rs201983499 Chr.X: 49174330-49174330 on Build GRCh38
rs782564725 Chr.X: 49174341-49174341 on Build GRCh38
rs374629732 Chr.X: 49174293-49174293 on Build GRCh38
rs957390148 Chr.X: 49174303-49174303 on Build GRCh38
rs371982481 Chr.X: 49174403-49174403 on Build GRCh38
rs781979194 Chr.X: 49174467-49174468 on Build GRCh38
rs782067756 Chr.X: 49174465-49174465 on Build GRCh38
rs782315999 Chr.X: 49174408-49174408 on Build GRCh38
rs782695122 Chr.X: 49174349-49174349 on Build GRCh38
rs782426022 Chr.X: 49174318-49174318 on Build GRCh38
rs782360262 Chr.X: 49174296-49174296 on Build GRCh38
rs376263784 Chr.X: 49174413-49174413 on Build GRCh38
rs922164426 Chr.X: 49174420-49174420 on Build GRCh38
rs782634210 Chr.X: 49174405-49174405 on Build GRCh38
rs781796769 Chr.X: 49174429-49174430 on Build GRCh38
rs782798398 Chr.X: 49174388-49174388 on Build GRCh38
rs11539258 Chr.X: 49174354-49174354 on Build GRCh38
rs782427974 Chr.X: 49174457-49174458 on Build GRCh38
rs782624428 Chr.X: 49174392-49174392 on Build GRCh38
rs782360280 Chr.X: 49174351-49174351 on Build GRCh38
rs782265454 Chr.X: 49174395-49174395 on Build GRCh38
rs782739679 Chr.X: 49174484-49174484 on Build GRCh38
rs375975539 Chr.X: 49174355-49174355 on Build GRCh38
rs781961016 Chr.X: 49174312-49174312 on Build GRCh38
rs782545835 Chr.X: 49174361-49174361 on Build GRCh38
rs782440793 Chr.X: 49174391-49174391 on Build GRCh38

View Full Product Details