Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28738114-28738339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEK8
Entrez Gene ID
284086
Gene Name
NIMA related kinase 8
Gene Aliases
JCK, NEK12A, NPHP9, RHPD2
UniGene
Hs.448468
Gene Chromosome Location
Chr.17: 28727899-28743457 on Build GRCh38
Transcripts
NM_178170.2 XM_011524638.2 XM_011524640.2 XM_017024499.1 XM_017024500.1 XM_017024501.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3717264 c.1100G>A p.S367N Chr.17: 28738123-28738123 on Build GRCh38
6146382 c.1184A>T p.H395L Chr.17: 28738207-28738207 on Build GRCh38
4459006 c.1108C>T p.P370S Chr.17: 28738131-28738131 on Build GRCh38
5346525 c.1156G>A p.E386K Chr.17: 28738179-28738179 on Build GRCh38
5346526 c.1173G>A p.V391V Chr.17: 28738196-28738196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769454263 Chr.17: 28738251-28738251 on Build GRCh38
rs375613242 Chr.17: 28738256-28738256 on Build GRCh38
rs374930363 Chr.17: 28738204-28738204 on Build GRCh38
rs767422789 Chr.17: 28738216-28738216 on Build GRCh38
rs138066977 Chr.17: 28738171-28738171 on Build GRCh38
rs757916936 Chr.17: 28738137-28738137 on Build GRCh38
rs367654888 Chr.17: 28738136-28738136 on Build GRCh38
rs772333473 Chr.17: 28738262-28738262 on Build GRCh38
rs368061384 Chr.17: 28738255-28738255 on Build GRCh38
rs755900226 Chr.17: 28738221-28738221 on Build GRCh38
rs773902843 Chr.17: 28738190-28738190 on Build GRCh38
rs371406871 Chr.17: 28738200-28738200 on Build GRCh38
rs574347342 Chr.17: 28738208-28738208 on Build GRCh38
rs777152445 Chr.17: 28738141-28738141 on Build GRCh38
rs763610690 Chr.17: 28738206-28738206 on Build GRCh38
rs199853008 Chr.17: 28738189-28738189 on Build GRCh38
rs771364513 Chr.17: 28738280-28738280 on Build GRCh38
rs186958324 Chr.17: 28738223-28738223 on Build GRCh38
rs780916071 Chr.17: 28738171-28738172 on Build GRCh38
rs145862009 Chr.17: 28738209-28738209 on Build GRCh38
rs747712613 Chr.17: 28738234-28738235 on Build GRCh38
rs748643938 Chr.17: 28738183-28738183 on Build GRCh38
rs749473321 Chr.17: 28738243-28738243 on Build GRCh38
rs146989750 Chr.17: 28738168-28738168 on Build GRCh38
rs191806152 Chr.17: 28738276-28738276 on Build GRCh38
rs906602387 Chr.17: 28738257-28738257 on Build GRCh38
rs779830257 Chr.17: 28738239-28738239 on Build GRCh38
rs775138270 Chr.17: 28738202-28738202 on Build GRCh38
rs750419550 Chr.17: 28738220-28738220 on Build GRCh38
rs751455589 Chr.17: 28738143-28738143 on Build GRCh38
rs757806007 Chr.17: 28738270-28738270 on Build GRCh38
rs780241960 Chr.17: 28738170-28738170 on Build GRCh38
rs780922648 Chr.17: 28738169-28738169 on Build GRCh38
rs3809797 Chr.17: 28738193-28738193 on Build GRCh38
rs773847705 Chr.17: 28738269-28738269 on Build GRCh38

View Full Product Details