Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31623533-31623806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PISD
Entrez Gene ID
23761
Gene Name
phosphatidylserine decarboxylase
Gene Aliases
DJ858B16, PSD, PSDC, PSSC, dJ858B16.2
UniGene
Hs.420559
Gene Chromosome Location
Chr.22: 31618491-31630824 on Build GRCh38
Transcripts
NM_001326411.1 NM_001326412.1 NM_001326413.1 NM_001326414.1 NM_001326415.1 NM_001326416.1 NM_001326417.1 NM_001326418.1 NM_001326419.1 NM_001326420.1 NM_001326421.1 NM_014338.3 NM_178022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6300617 c.211G>C p.V71L Chr.22: 31623692-31623692 on Build GRCh38
7416750 c.178C>A p.L60I Chr.22: 31623725-31623725 on Build GRCh38
4459063 c.110C>T p.S37F Chr.22: 31623793-31623793 on Build GRCh38
2938325 c.175G>A p.A59T Chr.22: 31623728-31623728 on Build GRCh38
444902 c.99G>A p.L33L Chr.22: 31623804-31623804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749185587 Chr.22: 31623728-31623728 on Build GRCh38
rs953049577 Chr.22: 31623652-31623652 on Build GRCh38
rs141299772 Chr.22: 31623718-31623718 on Build GRCh38
rs200329463 Chr.22: 31623644-31623644 on Build GRCh38
rs764242570 Chr.22: 31623772-31623772 on Build GRCh38
rs761331516 Chr.22: 31623689-31623689 on Build GRCh38
rs750648956 Chr.22: 31623785-31623785 on Build GRCh38
rs777669105 Chr.22: 31623705-31623705 on Build GRCh38
rs773668209 Chr.22: 31623735-31623735 on Build GRCh38
rs751842641 Chr.22: 31623709-31623709 on Build GRCh38
rs758469811 Chr.22: 31623651-31623651 on Build GRCh38
rs753171358 Chr.22: 31623695-31623695 on Build GRCh38
rs752730292 Chr.22: 31623776-31623777 on Build GRCh38
rs778097770 Chr.22: 31623655-31623655 on Build GRCh38
rs767669737 Chr.22: 31623783-31623783 on Build GRCh38
rs57725422 Chr.22: 31623710-31623710 on Build GRCh38
rs376939663 Chr.22: 31623559-31623559 on Build GRCh38
rs111934124 Chr.22: 31623555-31623555 on Build GRCh38
rs759936162 Chr.22: 31623756-31623756 on Build GRCh38
rs893000974 Chr.22: 31623560-31623560 on Build GRCh38
rs778975691 Chr.22: 31623643-31623643 on Build GRCh38
rs758522926 Chr.22: 31623696-31623696 on Build GRCh38
rs749241259 Chr.22: 31623787-31623787 on Build GRCh38
rs781057071 Chr.22: 31623714-31623714 on Build GRCh38
rs996702864 Chr.22: 31623593-31623593 on Build GRCh38
rs765563551 Chr.22: 31623764-31623764 on Build GRCh38
rs201519936 Chr.22: 31623636-31623636 on Build GRCh38
rs763333149 Chr.22: 31623770-31623770 on Build GRCh38
rs774074651 Chr.22: 31623688-31623688 on Build GRCh38
rs766769743 Chr.22: 31623690-31623690 on Build GRCh38
rs780185289 Chr.22: 31623726-31623726 on Build GRCh38
rs768025970 Chr.22: 31623678-31623678 on Build GRCh38
rs747177542 Chr.22: 31623656-31623656 on Build GRCh38
rs775561013 Chr.22: 31623670-31623670 on Build GRCh38
rs183406976 Chr.22: 31623755-31623755 on Build GRCh38
rs768243917 Chr.22: 31623731-31623731 on Build GRCh38
rs377226392 Chr.22: 31623681-31623681 on Build GRCh38
rs1028174666 Chr.22: 31623596-31623596 on Build GRCh38
rs770786006 Chr.22: 31623657-31623657 on Build GRCh38
rs199783760 Chr.22: 31623659-31623659 on Build GRCh38
rs745807486 Chr.22: 31623668-31623668 on Build GRCh38
rs146960059 Chr.22: 31623750-31623750 on Build GRCh38
rs1041251306 Chr.22: 31623600-31623600 on Build GRCh38
rs149109296 Chr.22: 31623692-31623692 on Build GRCh38
rs901300788 Chr.22: 31623635-31623635 on Build GRCh38
rs762588576 Chr.22: 31623677-31623677 on Build GRCh38
rs891057125 Chr.22: 31623599-31623599 on Build GRCh38
rs1049558978 Chr.22: 31623771-31623771 on Build GRCh38
rs1008276687 Chr.22: 31623637-31623637 on Build GRCh38
rs973881750 Chr.22: 31623662-31623662 on Build GRCh38
rs1054688615 Chr.22: 31623556-31623556 on Build GRCh38
rs751538096 Chr.22: 31623773-31623773 on Build GRCh38
rs765802509 Chr.22: 31623693-31623693 on Build GRCh38
rs931327080 Chr.22: 31623575-31623575 on Build GRCh38
rs757349016 Chr.22: 31623782-31623782 on Build GRCh38
rs940285871 Chr.22: 31623729-31623729 on Build GRCh38
rs148252495 Chr.22: 31623717-31623717 on Build GRCh38
rs137966413 Chr.22: 31623786-31623786 on Build GRCh38
rs887353700 Chr.22: 31623597-31623597 on Build GRCh38
rs771678477 Chr.22: 31623753-31623753 on Build GRCh38
rs776000864 Chr.22: 31623769-31623769 on Build GRCh38

View Full Product Details