Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 22982013-22982272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP8
Entrez Gene ID
5932
Gene Name
RB binding protein 8, endonuclease
Gene Aliases
COM1, CTIP, JWDS, RIM, SAE2, SCKL2
UniGene
Hs.546282
Gene Chromosome Location
Chr.18: 22914121-23026486 on Build GRCh38
Transcripts
NM_002894.2 NM_203291.1 NM_203292.1 XM_005258325.2 XM_005258326.3 XM_006722519.2 XM_006722520.2 XM_006722521.2 XM_011526132.2 XM_017025915.1 XM_017025916.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6465330 c.451G>A p.A151T Chr.18: 22982240-22982240 on Build GRCh38
987008 c.475G>A p.V159I Chr.18: 22982264-22982264 on Build GRCh38
4805211 c.440A>G p.Q147R Chr.18: 22982229-22982229 on Build GRCh38
987009 c.475G>A p.V159I Chr.18: 22982264-22982264 on Build GRCh38
6465331 c.451G>A p.A151T Chr.18: 22982240-22982240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755077339 Chr.18: 22982198-22982198 on Build GRCh38
rs776544939 Chr.18: 22982179-22982179 on Build GRCh38
rs754310666 Chr.18: 22982194-22982194 on Build GRCh38
rs139016995 Chr.18: 22982188-22982188 on Build GRCh38
rs114736611 Chr.18: 22982174-22982174 on Build GRCh38
rs959326509 Chr.18: 22982060-22982060 on Build GRCh38
rs199948715 Chr.18: 22982202-22982202 on Build GRCh38
rs373954478 Chr.18: 22982168-22982168 on Build GRCh38
rs567193499 Chr.18: 22982067-22982067 on Build GRCh38
rs758647514 Chr.18: 22982210-22982210 on Build GRCh38
rs935108954 Chr.18: 22982099-22982099 on Build GRCh38
rs977902252 Chr.18: 22982186-22982186 on Build GRCh38
rs538343074 Chr.18: 22982211-22982211 on Build GRCh38
rs769686702 Chr.18: 22982232-22982232 on Build GRCh38
rs749228733 Chr.18: 22982246-22982246 on Build GRCh38
rs756423611 Chr.18: 22982182-22982182 on Build GRCh38
rs181951749 Chr.18: 22982056-22982056 on Build GRCh38
rs368670826 Chr.18: 22982235-22982235 on Build GRCh38
rs745678667 Chr.18: 22982214-22982214 on Build GRCh38
rs750817155 Chr.18: 22982180-22982180 on Build GRCh38
rs536069092 Chr.18: 22982123-22982123 on Build GRCh38
rs748117894 Chr.18: 22982206-22982206 on Build GRCh38
rs1024887948 Chr.18: 22982081-22982081 on Build GRCh38

View Full Product Details