Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13164100-13164343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCM10
Entrez Gene ID
55388
Gene Name
minichromosome maintenance 10 replication initiation factor
Gene Aliases
CNA43, DNA43, PRO2249
UniGene
Hs.198363
Gene Chromosome Location
Chr.10: 13161554-13211110 on Build GRCh38
Transcripts
NM_018518.4 NM_182751.2 XM_011519538.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6302496 c.3G>T p.M1I Chr.10: 13164205-13164205 on Build GRCh38
6302497 c.3G>T p.M1I Chr.10: 13164205-13164205 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1006593870 Chr.10: 13164263-13164263 on Build GRCh38
rs1042228443 Chr.10: 13164300-13164300 on Build GRCh38
rs775017690 Chr.10: 13164251-13164251 on Build GRCh38
rs779007412 Chr.10: 13164194-13164194 on Build GRCh38
rs977926039 Chr.10: 13164316-13164316 on Build GRCh38
rs57536303 Chr.10: 13164296-13164296 on Build GRCh38
rs752939959 Chr.10: 13164221-13164221 on Build GRCh38
rs773180981 Chr.10: 13164217-13164217 on Build GRCh38
rs750764014 Chr.10: 13164162-13164162 on Build GRCh38
rs907327181 Chr.10: 13164138-13164138 on Build GRCh38
rs771048107 Chr.10: 13164236-13164236 on Build GRCh38
rs767891616 Chr.10: 13164154-13164154 on Build GRCh38
rs577060555 Chr.10: 13164274-13164274 on Build GRCh38
rs377614185 Chr.10: 13164270-13164270 on Build GRCh38
rs780538249 Chr.10: 13164170-13164170 on Build GRCh38
rs115861395 Chr.10: 13164169-13164169 on Build GRCh38
rs776444959 Chr.10: 13164238-13164238 on Build GRCh38
rs749445077 Chr.10: 13164179-13164179 on Build GRCh38
rs750857718 Chr.10: 13164261-13164261 on Build GRCh38
rs945421064 Chr.10: 13164299-13164299 on Build GRCh38
rs765463192 Chr.10: 13164143-13164143 on Build GRCh38
rs762643819 Chr.10: 13164254-13164254 on Build GRCh38
rs763709396 Chr.10: 13164256-13164256 on Build GRCh38
rs772397812 Chr.10: 13164216-13164216 on Build GRCh38
rs748336688 Chr.10: 13164207-13164207 on Build GRCh38
rs961392602 Chr.10: 13164133-13164133 on Build GRCh38
rs755015664 Chr.10: 13164187-13164187 on Build GRCh38
rs7090944 Chr.10: 13164233-13164233 on Build GRCh38
rs373360430 Chr.10: 13164242-13164242 on Build GRCh38
rs557127753 Chr.10: 13164244-13164244 on Build GRCh38

View Full Product Details