Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 75647055-75647302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL19
Entrez Gene ID
9801
Gene Name
mitochondrial ribosomal protein L19
Gene Aliases
L19mt, MRP-L15, MRP-L19, MRPL15, RLX1, RPML15
UniGene
Hs.44024
Gene Chromosome Location
Chr.2: 75646783-75662208 on Build GRCh38
Transcripts
NM_014763.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5496060 c.158C>T p.A53V Chr.2: 75647156-75647156 on Build GRCh38
5496061 c.167C>T p.P56L Chr.2: 75647165-75647165 on Build GRCh38
6715289 c.143C>A p.P48H Chr.2: 75647141-75647141 on Build GRCh38
4769683 c.221+9C>T p.? Chr.2: 75647228-75647228 on Build GRCh38
722449 c.218G>A p.R73H Chr.2: 75647216-75647216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564694099 Chr.2: 75647229-75647229 on Build GRCh38
rs780776529 Chr.2: 75647165-75647165 on Build GRCh38
rs779651858 Chr.2: 75647179-75647179 on Build GRCh38
rs779415298 Chr.2: 75647240-75647240 on Build GRCh38
rs1026715782 Chr.2: 75647239-75647239 on Build GRCh38
rs1054707255 Chr.2: 75647245-75647245 on Build GRCh38
rs528861522 Chr.2: 75647276-75647276 on Build GRCh38
rs768411635 Chr.2: 75647193-75647193 on Build GRCh38
rs778639577 Chr.2: 75647247-75647247 on Build GRCh38
rs747979715 Chr.2: 75647202-75647202 on Build GRCh38
rs755713132 Chr.2: 75647167-75647167 on Build GRCh38
rs768094286 Chr.2: 75647236-75647236 on Build GRCh38
rs756508201 Chr.2: 75647260-75647267 on Build GRCh38
rs758688463 Chr.2: 75647116-75647116 on Build GRCh38
rs753452749 Chr.2: 75647242-75647242 on Build GRCh38
rs370208064 Chr.2: 75647216-75647216 on Build GRCh38
rs773155948 Chr.2: 75647205-75647205 on Build GRCh38
rs751808358 Chr.2: 75647232-75647232 on Build GRCh38
rs750926785 Chr.2: 75647237-75647237 on Build GRCh38
rs546128151 Chr.2: 75647228-75647228 on Build GRCh38
rs35821342 Chr.2: 75647111-75647112 on Build GRCh38
rs200706958 Chr.2: 75647238-75647238 on Build GRCh38
rs1001211850 Chr.2: 75647241-75647241 on Build GRCh38
rs771886271 Chr.2: 75647265-75647265 on Build GRCh38
rs2286239 Chr.2: 75647264-75647264 on Build GRCh38
rs546630780 Chr.2: 75647244-75647244 on Build GRCh38
rs190250599 Chr.2: 75647083-75647083 on Build GRCh38
rs757839957 Chr.2: 75647149-75647149 on Build GRCh38
rs1048045864 Chr.2: 75647137-75647137 on Build GRCh38
rs373641389 Chr.2: 75647204-75647204 on Build GRCh38
rs1013900637 Chr.2: 75647166-75647166 on Build GRCh38
rs1045505355 Chr.2: 75647212-75647212 on Build GRCh38
rs781766726 Chr.2: 75647157-75647157 on Build GRCh38
rs182071884 Chr.2: 75647084-75647084 on Build GRCh38
rs575511320 Chr.2: 75647201-75647201 on Build GRCh38
rs777586698 Chr.2: 75647271-75647271 on Build GRCh38
rs752896158 Chr.2: 75647108-75647108 on Build GRCh38
rs765395227 Chr.2: 75647098-75647098 on Build GRCh38
rs1024923447 Chr.2: 75647170-75647170 on Build GRCh38
rs920519649 Chr.2: 75647119-75647119 on Build GRCh38
rs369807553 Chr.2: 75647081-75647081 on Build GRCh38
rs759627454 Chr.2: 75647090-75647090 on Build GRCh38
rs542131299 Chr.2: 75647091-75647091 on Build GRCh38
rs892168917 Chr.2: 75647154-75647154 on Build GRCh38
rs867740619 Chr.2: 75647171-75647171 on Build GRCh38
rs904997301 Chr.2: 75647180-75647180 on Build GRCh38
rs377418255 Chr.2: 75647209-75647209 on Build GRCh38
rs762286179 Chr.2: 75647234-75647234 on Build GRCh38
rs907605616 Chr.2: 75647206-75647206 on Build GRCh38
rs775688980 Chr.2: 75647224-75647224 on Build GRCh38
rs748866193 Chr.2: 75647192-75647192 on Build GRCh38
rs753263546 Chr.2: 75647176-75647176 on Build GRCh38
rs930476214 Chr.2: 75647135-75647135 on Build GRCh38
rs987654598 Chr.2: 75647252-75647252 on Build GRCh38
rs978909276 Chr.2: 75647273-75647273 on Build GRCh38
rs910824969 Chr.2: 75647253-75647253 on Build GRCh38
rs201335545 Chr.2: 75647146-75647146 on Build GRCh38

View Full Product Details