Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72227864-72228131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDA
Entrez Gene ID
9615
Gene Name
guanine deaminase
Gene Aliases
CYPIN, GUANASE, NEDASIN
UniGene
Hs.494163
Gene Chromosome Location
Chr.9: 72114595-72256045 on Build GRCh38
Transcripts
NM_001242505.2 NM_001242506.2 NM_001242507.2 NM_004293.4 XM_005252317.1 XM_011519213.1 XM_011519214.1 XM_011519215.1 XM_011519216.1 XM_011519217.1 XM_011519218.2 XM_011519219.1 XM_011519220.1 XM_011519221.1 XM_011519222.1 XM_011519223.2 XM_017015333.1 XM_017015334.1 XM_017015335.1 XM_017015336.1 XM_017015337.1 XM_017015338.1 XM_017015339.1 XM_017015340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7146852 c.837C>T p.H279H Chr.9: 72227957-72227957 on Build GRCh38
5027076 c.475G>A p.E159K Chr.9: 72227997-72227997 on Build GRCh38
7096915 c.849C>T p.L283L Chr.9: 72227969-72227969 on Build GRCh38
4567810 c.883_884GG>AA p.G295K Chr.9: 72228003-72228004 on Build GRCh38
7146853 c.435C>T p.H145H Chr.9: 72227957-72227957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143236383 Chr.9: 72228001-72228001 on Build GRCh38
rs974947363 Chr.9: 72228072-72228072 on Build GRCh38
rs950236644 Chr.9: 72227914-72227914 on Build GRCh38
rs369386185 Chr.9: 72228077-72228077 on Build GRCh38
rs371570414 Chr.9: 72227995-72227995 on Build GRCh38
rs901382579 Chr.9: 72227915-72227915 on Build GRCh38
rs754744003 Chr.9: 72228011-72228011 on Build GRCh38
rs375884548 Chr.9: 72228071-72228071 on Build GRCh38
rs148310315 Chr.9: 72228028-72228028 on Build GRCh38
rs17579467 Chr.9: 72227935-72227935 on Build GRCh38
rs773396126 Chr.9: 72227964-72227964 on Build GRCh38
rs547662878 Chr.9: 72228012-72228012 on Build GRCh38
rs749376626 Chr.9: 72228034-72228034 on Build GRCh38
rs758297575 Chr.9: 72227921-72227921 on Build GRCh38
rs200060290 Chr.9: 72227909-72227909 on Build GRCh38
rs764854501 Chr.9: 72228078-72228078 on Build GRCh38
rs567468915 Chr.9: 72228064-72228064 on Build GRCh38
rs756519737 Chr.9: 72227893-72227893 on Build GRCh38
rs372389322 Chr.9: 72227940-72227940 on Build GRCh38
rs756736155 Chr.9: 72228052-72228052 on Build GRCh38
rs374053200 Chr.9: 72228061-72228061 on Build GRCh38
rs1033061194 Chr.9: 72227973-72227973 on Build GRCh38
rs1028827725 Chr.9: 72228092-72228092 on Build GRCh38
rs763605798 Chr.9: 72227967-72227967 on Build GRCh38
rs746792585 Chr.9: 72227925-72227925 on Build GRCh38
rs778734702 Chr.9: 72227910-72227910 on Build GRCh38
rs148910433 Chr.9: 72227985-72227985 on Build GRCh38
rs777479638 Chr.9: 72227904-72227904 on Build GRCh38
rs148533523 Chr.9: 72228004-72228004 on Build GRCh38
rs369889349 Chr.9: 72228049-72228049 on Build GRCh38
rs371594886 Chr.9: 72227922-72227922 on Build GRCh38
rs773485893 Chr.9: 72228016-72228016 on Build GRCh38
rs771557496 Chr.9: 72228084-72228101 on Build GRCh38
rs745761286 Chr.9: 72227920-72227920 on Build GRCh38
rs748443792 Chr.9: 72228015-72228015 on Build GRCh38
rs749764388 Chr.9: 72228040-72228040 on Build GRCh38
rs756409262 Chr.9: 72227956-72227956 on Build GRCh38
rs551530651 Chr.9: 72227958-72227958 on Build GRCh38
rs375919938 Chr.9: 72228007-72228007 on Build GRCh38
rs780378291 Chr.9: 72228096-72228096 on Build GRCh38
rs771573154 Chr.9: 72228041-72228041 on Build GRCh38
rs761246444 Chr.9: 72228029-72228029 on Build GRCh38
rs189759518 Chr.9: 72227972-72227972 on Build GRCh38
rs999603417 Chr.9: 72227990-72227990 on Build GRCh38
rs368196362 Chr.9: 72227939-72227939 on Build GRCh38
rs753477191 Chr.9: 72227987-72227987 on Build GRCh38
rs758097286 Chr.9: 72227908-72227908 on Build GRCh38
rs368033130 Chr.9: 72227988-72227988 on Build GRCh38
rs748189548 Chr.9: 72227957-72227957 on Build GRCh38
rs759881702 Chr.9: 72227986-72227986 on Build GRCh38
rs746811009 Chr.9: 72228008-72228008 on Build GRCh38

View Full Product Details