Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50531932-50532165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ODF3B
Entrez Gene ID
440836
Gene Name
outer dense fiber of sperm tails 3B
Gene Aliases
ODF3L3
UniGene
Hs.531314
Gene Chromosome Location
Chr.22: 50530409-50532579 on Build GRCh38
Transcripts
NM_001014440.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4624761 c.48C>T p.R16R Chr.22: 50532030-50532030 on Build GRCh38
4650607 c.37C>T p.H13Y Chr.22: 50532041-50532041 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs540657372 Chr.22: 50531980-50531980 on Build GRCh38
rs1006008137 Chr.22: 50531960-50531960 on Build GRCh38
rs868639633 Chr.22: 50532008-50532008 on Build GRCh38
rs923306993 Chr.22: 50532113-50532113 on Build GRCh38
rs910509391 Chr.22: 50532134-50532135 on Build GRCh38
rs933310218 Chr.22: 50532131-50532131 on Build GRCh38
rs962000214 Chr.22: 50532030-50532030 on Build GRCh38
rs866693845 Chr.22: 50531969-50531969 on Build GRCh38
rs12169889 Chr.22: 50532087-50532087 on Build GRCh38
rs761678368 Chr.22: 50532012-50532012 on Build GRCh38
rs751126160 Chr.22: 50532068-50532068 on Build GRCh38
rs993797270 Chr.22: 50532046-50532046 on Build GRCh38
rs866079670 Chr.22: 50531996-50531996 on Build GRCh38
rs529965622 Chr.22: 50532081-50532081 on Build GRCh38
rs376010265 Chr.22: 50532016-50532016 on Build GRCh38
rs768385726 Chr.22: 50532124-50532124 on Build GRCh38
rs901963288 Chr.22: 50531957-50531957 on Build GRCh38
rs750117853 Chr.22: 50532017-50532017 on Build GRCh38
rs1033423396 Chr.22: 50532065-50532065 on Build GRCh38
rs765993054 Chr.22: 50532061-50532061 on Build GRCh38
rs573269147 Chr.22: 50531962-50531962 on Build GRCh38
rs970711215 Chr.22: 50532036-50532036 on Build GRCh38
rs778039775 Chr.22: 50532094-50532094 on Build GRCh38
rs1050510708 Chr.22: 50532137-50532137 on Build GRCh38
rs772116713 Chr.22: 50532004-50532004 on Build GRCh38
rs781675513 Chr.22: 50532083-50532083 on Build GRCh38
rs771248121 Chr.22: 50531961-50531961 on Build GRCh38
rs946372342 Chr.22: 50532093-50532093 on Build GRCh38
rs978039887 Chr.22: 50532105-50532105 on Build GRCh38
rs865981361 Chr.22: 50532014-50532014 on Build GRCh38
rs559668137 Chr.22: 50532021-50532021 on Build GRCh38
rs867736412 Chr.22: 50532019-50532019 on Build GRCh38
rs371513060 Chr.22: 50532088-50532088 on Build GRCh38
rs764196896 Chr.22: 50531987-50531987 on Build GRCh38
rs1006891913 Chr.22: 50532001-50532001 on Build GRCh38
rs754478259 Chr.22: 50532076-50532076 on Build GRCh38
rs980548858 Chr.22: 50532064-50532064 on Build GRCh38
rs1024645007 Chr.22: 50532054-50532054 on Build GRCh38

View Full Product Details