Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46203396-46203663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LURAP1
Entrez Gene ID
541468
Gene Name
leucine rich adaptor protein 1
Gene Aliases
C1orf190, LRAP35a, LRP35A
UniGene
Hs.568642
Gene Chromosome Location
Chr.1: 46203334-46221256 on Build GRCh38
Transcripts
NM_001013615.2 NM_001243766.1

Gene Symbol
POMGNT1
Entrez Gene ID
55624
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-)
Gene Aliases
GNTI.2, GnT I.2, LGMD2O, MEB, MGAT1.2, gnT-I.2
UniGene
Hs.525134
Gene Chromosome Location
Chr.1: 46188681-46220305 on Build GRCh38
Transcripts
NM_001013615.2 NM_001243766.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5448682 c.99C>T p.G33G Chr.1: 46203525-46203525 on Build GRCh38
5763827 c.144G>A p.A48A Chr.1: 46203570-46203570 on Build GRCh38
5992624 c.198+2T>C p.? Chr.1: 46203626-46203626 on Build GRCh38
7191573 c.121G>A p.G41S Chr.1: 46203547-46203547 on Build GRCh38
3689740 c.166G>A p.G56R Chr.1: 46203592-46203592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552194278 Chr.1: 46203564-46203564 on Build GRCh38
rs200199469 Chr.1: 46203553-46203553 on Build GRCh38
rs776919489 Chr.1: 46203465-46203465 on Build GRCh38
rs761422074 Chr.1: 46203503-46203503 on Build GRCh38
rs571957976 Chr.1: 46203424-46203424 on Build GRCh38
rs888785493 Chr.1: 46203457-46203457 on Build GRCh38
rs773947920 Chr.1: 46203499-46203499 on Build GRCh38
rs1009384532 Chr.1: 46203539-46203539 on Build GRCh38
rs773440254 Chr.1: 46203433-46203433 on Build GRCh38
rs760341311 Chr.1: 46203632-46203632 on Build GRCh38
rs549837624 Chr.1: 46203616-46203616 on Build GRCh38
rs772894417 Chr.1: 46203621-46203621 on Build GRCh38
rs909021290 Chr.1: 46203547-46203547 on Build GRCh38
rs967335548 Chr.1: 46203462-46203462 on Build GRCh38
rs570474263 Chr.1: 46203600-46203600 on Build GRCh38
rs748260520 Chr.1: 46203423-46203423 on Build GRCh38
rs377472143 Chr.1: 46203577-46203577 on Build GRCh38
rs747307558 Chr.1: 46203452-46203452 on Build GRCh38
rs144228893 Chr.1: 46203521-46203521 on Build GRCh38
rs1051424782 Chr.1: 46203426-46203426 on Build GRCh38
rs562545438 Chr.1: 46203510-46203510 on Build GRCh38
rs746239517 Chr.1: 46203589-46203589 on Build GRCh38
rs767211043 Chr.1: 46203504-46203504 on Build GRCh38
rs199661717 Chr.1: 46203559-46203559 on Build GRCh38
rs62620991 Chr.1: 46203604-46203604 on Build GRCh38
rs959379326 Chr.1: 46203609-46203609 on Build GRCh38
rs766172760 Chr.1: 46203513-46203513 on Build GRCh38
rs200435573 Chr.1: 46203569-46203569 on Build GRCh38
rs1020548799 Chr.1: 46203428-46203428 on Build GRCh38
rs760067527 Chr.1: 46203476-46203476 on Build GRCh38
rs992011525 Chr.1: 46203543-46203543 on Build GRCh38
rs771110165 Chr.1: 46203580-46203580 on Build GRCh38
rs958086754 Chr.1: 46203534-46203534 on Build GRCh38
rs765693469 Chr.1: 46203479-46203479 on Build GRCh38
rs771323153 Chr.1: 46203453-46203453 on Build GRCh38
rs142875865 Chr.1: 46203550-46203550 on Build GRCh38
rs759582353 Chr.1: 46203595-46203595 on Build GRCh38
rs984591393 Chr.1: 46203622-46203622 on Build GRCh38
rs772237850 Chr.1: 46203425-46203425 on Build GRCh38
rs759465370 Chr.1: 46203464-46203464 on Build GRCh38
rs981456825 Chr.1: 46203485-46203485 on Build GRCh38
rs750092774 Chr.1: 46203507-46203507 on Build GRCh38

View Full Product Details