Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88734407-88734658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100289580
Entrez Gene ID
100289580
Gene Name
uncharacterized LOC100289580
Gene Aliases
-
UniGene
Hs.437712
Gene Chromosome Location
Chr.16: 88731180-88741425 on Build GRCh38
Transcripts
NR_103774.1 NM_001142864.3

Gene Symbol
PIEZO1
Entrez Gene ID
9780
Gene Name
piezo type mechanosensitive ion channel component 1
Gene Aliases
DHS, FAM38A, LMPH3, Mib
UniGene
Hs.377001
Gene Chromosome Location
Chr.16: 88715338-88785220 on Build GRCh38
Transcripts
NR_103774.1 NM_001142864.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3755217 c.2031G>A p.V677V Chr.16: 88734505-88734505 on Build GRCh38
6377279 c.2048G>A p.S683N Chr.16: 88734488-88734488 on Build GRCh38
4632137 c.2045C>T p.S682F Chr.16: 88734491-88734491 on Build GRCh38
5523468 c.2046C>T p.S682S Chr.16: 88734490-88734490 on Build GRCh38
4476240 c.2052C>T p.I684I Chr.16: 88734484-88734484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552433164 Chr.16: 88734563-88734563 on Build GRCh38
rs541569160 Chr.16: 88734541-88734541 on Build GRCh38
rs562438565 Chr.16: 88734507-88734507 on Build GRCh38
rs568566636 Chr.16: 88734604-88734604 on Build GRCh38
rs1015932479 Chr.16: 88734597-88734597 on Build GRCh38
rs942475590 Chr.16: 88734625-88734625 on Build GRCh38
rs761977673 Chr.16: 88734632-88734632 on Build GRCh38
rs766101007 Chr.16: 88734580-88734580 on Build GRCh38
rs910070855 Chr.16: 88734598-88734598 on Build GRCh38
rs370541429 Chr.16: 88734508-88734508 on Build GRCh38
rs79613370 Chr.16: 88734594-88734594 on Build GRCh38
rs781511572 Chr.16: 88734571-88734572 on Build GRCh38
rs918916882 Chr.16: 88734615-88734615 on Build GRCh38
rs933689569 Chr.16: 88734617-88734617 on Build GRCh38
rs188058617 Chr.16: 88734543-88734543 on Build GRCh38
rs375878215 Chr.16: 88734488-88734488 on Build GRCh38
rs746281782 Chr.16: 88734584-88734584 on Build GRCh38
rs1025224511 Chr.16: 88734454-88734454 on Build GRCh38
rs918800272 Chr.16: 88734502-88734502 on Build GRCh38
rs948942641 Chr.16: 88734447-88734447 on Build GRCh38
rs774632836 Chr.16: 88734631-88734631 on Build GRCh38
rs942831980 Chr.16: 88734634-88734634 on Build GRCh38
rs759891355 Chr.16: 88734436-88734436 on Build GRCh38
rs969196199 Chr.16: 88734474-88734474 on Build GRCh38
rs767599437 Chr.16: 88734633-88734633 on Build GRCh38
rs912747158 Chr.16: 88734628-88734628 on Build GRCh38
rs770217538 Chr.16: 88734608-88734608 on Build GRCh38
rs775681365 Chr.16: 88734611-88734611 on Build GRCh38
rs35742404 Chr.16: 88734475-88734475 on Build GRCh38
rs908903846 Chr.16: 88734481-88734481 on Build GRCh38
rs749533056 Chr.16: 88734614-88734614 on Build GRCh38
rs574426042 Chr.16: 88734532-88734532 on Build GRCh38
rs779660967 Chr.16: 88734593-88734593 on Build GRCh38
rs747598025 Chr.16: 88734568-88734568 on Build GRCh38
rs969606389 Chr.16: 88734579-88734579 on Build GRCh38
rs769950800 Chr.16: 88734472-88734472 on Build GRCh38
rs374761578 Chr.16: 88734606-88734606 on Build GRCh38
rs1024773651 Chr.16: 88734586-88734586 on Build GRCh38
rs889497081 Chr.16: 88734622-88734622 on Build GRCh38
rs751288031 Chr.16: 88734595-88734595 on Build GRCh38
rs886041515 Chr.16: 88734501-88734501 on Build GRCh38
rs3214056 Chr.16: 88734618-88734618 on Build GRCh38
rs777039661 Chr.16: 88734426-88734426 on Build GRCh38
rs564476931 Chr.16: 88734564-88734564 on Build GRCh38
rs531780121 Chr.16: 88734583-88734583 on Build GRCh38
rs891274876 Chr.16: 88734529-88734529 on Build GRCh38
rs34908386 Chr.16: 88734505-88734505 on Build GRCh38
rs1022134676 Chr.16: 88734552-88734552 on Build GRCh38
rs757829603 Chr.16: 88734570-88734570 on Build GRCh38
rs956075332 Chr.16: 88734575-88734575 on Build GRCh38
rs941576083 Chr.16: 88734607-88734607 on Build GRCh38
rs1007096231 Chr.16: 88734549-88734549 on Build GRCh38
rs917418405 Chr.16: 88734430-88734430 on Build GRCh38
rs1052352857 Chr.16: 88734561-88734561 on Build GRCh38
rs980300060 Chr.16: 88734462-88734462 on Build GRCh38
rs753550837 Chr.16: 88734562-88734562 on Build GRCh38
rs866496600 Chr.16: 88734473-88734473 on Build GRCh38
rs761259080 Chr.16: 88734496-88734496 on Build GRCh38
rs987439595 Chr.16: 88734626-88734626 on Build GRCh38
rs74033378 Chr.16: 88734548-88734548 on Build GRCh38

View Full Product Details