Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32836457-32836714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPRE1
Entrez Gene ID
22919
Gene Name
microtubule associated protein RP/EB family member 1
Gene Aliases
EB1
UniGene
Hs.472437
Gene Chromosome Location
Chr.20: 32819780-32850405 on Build GRCh38
Transcripts
NM_012325.2 XM_011528696.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6336593 c.330G>T p.W110C Chr.20: 32836696-32836696 on Build GRCh38
6307038 c.346G>T p.D116Y Chr.20: 32836712-32836712 on Build GRCh38
1130658 c.297A>G p.G99G Chr.20: 32836663-32836663 on Build GRCh38
3939302 c.345C>T p.F115F Chr.20: 32836711-32836711 on Build GRCh38
4700645 c.275C>A p.P92H Chr.20: 32836641-32836641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147611503 Chr.20: 32836657-32836657 on Build GRCh38
rs371042900 Chr.20: 32836688-32836688 on Build GRCh38
rs756925852 Chr.20: 32836596-32836596 on Build GRCh38
rs909794667 Chr.20: 32836561-32836561 on Build GRCh38
rs905632074 Chr.20: 32836684-32836684 on Build GRCh38
rs772573885 Chr.20: 32836599-32836600 on Build GRCh38
rs573827644 Chr.20: 32836610-32836610 on Build GRCh38
rs761353179 Chr.20: 32836634-32836634 on Build GRCh38
rs751154920 Chr.20: 32836562-32836562 on Build GRCh38
rs562058480 Chr.20: 32836483-32836483 on Build GRCh38
rs776602371 Chr.20: 32836595-32836595 on Build GRCh38
rs1044144737 Chr.20: 32836650-32836650 on Build GRCh38
rs754319096 Chr.20: 32836687-32836687 on Build GRCh38
rs564742924 Chr.20: 32836629-32836629 on Build GRCh38
rs775299849 Chr.20: 32836605-32836605 on Build GRCh38
rs768314433 Chr.20: 32836624-32836624 on Build GRCh38
rs774051033 Chr.20: 32836631-32836631 on Build GRCh38
rs769582976 Chr.20: 32836604-32836604 on Build GRCh38
rs766982774 Chr.20: 32836678-32836678 on Build GRCh38
rs113459843 Chr.20: 32836544-32836544 on Build GRCh38
rs186510921 Chr.20: 32836592-32836592 on Build GRCh38

View Full Product Details