Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46742864-46743135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS45
Entrez Gene ID
377047
Gene Name
protease, serine 45
Gene Aliases
TESSP5
UniGene
Hs.667712
Gene Chromosome Location
Chr.3: 46742091-46744755 on Build GRCh38
Transcripts
NM_199183.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3068372 c.413A>G p.D138G Chr.3: 46742953-46742953 on Build GRCh38
3760129 c.389C>T p.P130L Chr.3: 46742977-46742977 on Build GRCh38
7094794 c.352A>G p.R118G Chr.3: 46743014-46743014 on Build GRCh38
241273 c.362G>A p.R121Q Chr.3: 46743004-46743004 on Build GRCh38
1737542 c.331G>A p.V111M Chr.3: 46743035-46743035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766694070 Chr.3: 46742976-46742977 on Build GRCh38
rs773394844 Chr.3: 46742966-46742966 on Build GRCh38
rs372880035 Chr.3: 46742938-46742938 on Build GRCh38
rs756982801 Chr.3: 46743042-46743042 on Build GRCh38
rs773730494 Chr.3: 46742987-46742987 on Build GRCh38
rs747307974 Chr.3: 46742976-46742976 on Build GRCh38
rs62246251 Chr.3: 46743090-46743090 on Build GRCh38
rs58943210 Chr.3: 46742977-46742977 on Build GRCh38
rs371517365 Chr.3: 46743057-46743057 on Build GRCh38
rs141262874 Chr.3: 46743012-46743012 on Build GRCh38
rs766783908 Chr.3: 46743088-46743088 on Build GRCh38
rs750135794 Chr.3: 46743061-46743061 on Build GRCh38
rs369073318 Chr.3: 46742958-46742958 on Build GRCh38
rs773862766 Chr.3: 46743004-46743004 on Build GRCh38
rs375169812 Chr.3: 46742923-46742923 on Build GRCh38
rs751724295 Chr.3: 46743047-46743047 on Build GRCh38
rs375634062 Chr.3: 46742957-46742957 on Build GRCh38
rs762931884 Chr.3: 46742974-46742975 on Build GRCh38
rs755157722 Chr.3: 46743052-46743052 on Build GRCh38
rs767113456 Chr.3: 46742948-46742948 on Build GRCh38
rs753869543 Chr.3: 46743029-46743029 on Build GRCh38
rs768824478 Chr.3: 46742909-46742909 on Build GRCh38
rs763263445 Chr.3: 46743000-46743000 on Build GRCh38
rs753816580 Chr.3: 46743114-46743114 on Build GRCh38
rs556551810 Chr.3: 46742912-46742912 on Build GRCh38
rs764086811 Chr.3: 46743026-46743026 on Build GRCh38
rs751253561 Chr.3: 46742944-46742944 on Build GRCh38
rs372142899 Chr.3: 46742889-46742889 on Build GRCh38
rs78954493 Chr.3: 46743058-46743058 on Build GRCh38
rs748227553 Chr.3: 46742979-46742979 on Build GRCh38
rs762362943 Chr.3: 46742921-46742921 on Build GRCh38
rs778823255 Chr.3: 46743043-46743043 on Build GRCh38
rs780110977 Chr.3: 46742975-46742975 on Build GRCh38
rs367659910 Chr.3: 46743102-46743102 on Build GRCh38
rs759301679 Chr.3: 46743005-46743005 on Build GRCh38
rs932312887 Chr.3: 46742929-46742929 on Build GRCh38
rs759831600 Chr.3: 46743089-46743089 on Build GRCh38
rs1036767818 Chr.3: 46742980-46742980 on Build GRCh38
rs757153378 Chr.3: 46742972-46742972 on Build GRCh38
rs747022407 Chr.3: 46742893-46742893 on Build GRCh38
rs369099736 Chr.3: 46742932-46742932 on Build GRCh38
rs375360987 Chr.3: 46743091-46743091 on Build GRCh38
rs777866464 Chr.3: 46742961-46742961 on Build GRCh38
rs749046296 Chr.3: 46742966-46742966 on Build GRCh38
rs772366396 Chr.3: 46742885-46742885 on Build GRCh38
rs771366206 Chr.3: 46743002-46743002 on Build GRCh38
rs770224720 Chr.3: 46742982-46742982 on Build GRCh38
rs377053516 Chr.3: 46743014-46743014 on Build GRCh38
rs1000047144 Chr.3: 46743068-46743068 on Build GRCh38
rs377660279 Chr.3: 46743062-46743062 on Build GRCh38
rs548063198 Chr.3: 46743086-46743086 on Build GRCh38
rs749842005 Chr.3: 46743081-46743081 on Build GRCh38
rs771196370 Chr.3: 46743082-46743082 on Build GRCh38

View Full Product Details