Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70659495-70659760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL34
Entrez Gene ID
146433
Gene Name
interleukin 34
Gene Aliases
C16orf77, IL-34
UniGene
Hs.461214
Gene Chromosome Location
Chr.16: 70579895-70660682 on Build GRCh38
Transcripts
NM_001172771.1 NM_001172772.1 NM_152456.2 XM_011522902.2 XM_011522903.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5946338 c.530G>A p.C177Y Chr.16: 70659745-70659745 on Build GRCh38
3888835 c.403-1G>A p.? Chr.16: 70659617-70659617 on Build GRCh38
6398876 c.467T>C p.L156P Chr.16: 70659682-70659682 on Build GRCh38
4062387 c.470A>T p.K157M Chr.16: 70659685-70659685 on Build GRCh38
7089152 c.432C>A p.S144S Chr.16: 70659647-70659647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040421775 Chr.16: 70659527-70659528 on Build GRCh38
rs746397587 Chr.16: 70659606-70659606 on Build GRCh38
rs79339508 Chr.16: 70659520-70659520 on Build GRCh38
rs985414334 Chr.16: 70659732-70659732 on Build GRCh38
rs747817502 Chr.16: 70659696-70659696 on Build GRCh38
rs572757320 Chr.16: 70659670-70659670 on Build GRCh38
rs912744286 Chr.16: 70659530-70659530 on Build GRCh38
rs199550676 Chr.16: 70659575-70659575 on Build GRCh38
rs140718482 Chr.16: 70659638-70659638 on Build GRCh38
rs377763641 Chr.16: 70659585-70659585 on Build GRCh38
rs892086411 Chr.16: 70659561-70659561 on Build GRCh38
rs764948634 Chr.16: 70659600-70659601 on Build GRCh38
rs764258011 Chr.16: 70659667-70659667 on Build GRCh38
rs367851338 Chr.16: 70659724-70659724 on Build GRCh38
rs753344320 Chr.16: 70659693-70659693 on Build GRCh38
rs370863667 Chr.16: 70659695-70659695 on Build GRCh38
rs200891924 Chr.16: 70659657-70659657 on Build GRCh38
rs761630574 Chr.16: 70659596-70659599 on Build GRCh38
rs142214904 Chr.16: 70659723-70659723 on Build GRCh38
rs765228178 Chr.16: 70659662-70659662 on Build GRCh38
rs374146651 Chr.16: 70659681-70659681 on Build GRCh38
rs201478676 Chr.16: 70659647-70659647 on Build GRCh38
rs747915412 Chr.16: 70659641-70659641 on Build GRCh38
rs750885586 Chr.16: 70659680-70659680 on Build GRCh38
rs368367274 Chr.16: 70659694-70659694 on Build GRCh38
rs904092613 Chr.16: 70659563-70659563 on Build GRCh38
rs201108464 Chr.16: 70659666-70659666 on Build GRCh38
rs1014172148 Chr.16: 70659587-70659587 on Build GRCh38
rs773227906 Chr.16: 70659648-70659648 on Build GRCh38
rs775686925 Chr.16: 70659665-70659665 on Build GRCh38
rs932090107 Chr.16: 70659734-70659734 on Build GRCh38
rs141014593 Chr.16: 70659559-70659559 on Build GRCh38
rs546919435 Chr.16: 70659522-70659522 on Build GRCh38
rs866129766 Chr.16: 70659552-70659552 on Build GRCh38
rs755563995 Chr.16: 70659609-70659609 on Build GRCh38
rs571538473 Chr.16: 70659558-70659558 on Build GRCh38
rs369331265 Chr.16: 70659574-70659574 on Build GRCh38
rs774331963 Chr.16: 70659579-70659579 on Build GRCh38
rs944239924 Chr.16: 70659531-70659531 on Build GRCh38
rs1012233983 Chr.16: 70659566-70659566 on Build GRCh38
rs752849330 Chr.16: 70659591-70659591 on Build GRCh38
rs112537035 Chr.16: 70659601-70659601 on Build GRCh38
rs753727217 Chr.16: 70659592-70659592 on Build GRCh38
rs761824322 Chr.16: 70659580-70659580 on Build GRCh38
rs113364530 Chr.16: 70659581-70659581 on Build GRCh38
rs768802695 Chr.16: 70659600-70659600 on Build GRCh38
rs745340705 Chr.16: 70659572-70659572 on Build GRCh38
rs763997731 Chr.16: 70659571-70659572 on Build GRCh38
rs373785976 Chr.16: 70659592-70659592 on Build GRCh38
rs765373861 Chr.16: 70659586-70659586 on Build GRCh38
rs138898622 Chr.16: 70659677-70659677 on Build GRCh38
rs983346520 Chr.16: 70659655-70659655 on Build GRCh38
rs370570658 Chr.16: 70659637-70659637 on Build GRCh38
rs770853044 Chr.16: 70659729-70659729 on Build GRCh38
rs771982040 Chr.16: 70659710-70659710 on Build GRCh38
rs187166563 Chr.16: 70659673-70659673 on Build GRCh38

View Full Product Details