Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97666952-97667213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PI4K2A
Entrez Gene ID
55361
Gene Name
phosphatidylinositol 4-kinase type 2 alpha
Gene Aliases
PI4KII, PIK42A
UniGene
Hs.25300
Gene Chromosome Location
Chr.10: 97640686-97676434 on Build GRCh38
Transcripts
NM_018425.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7139601 c.1167G>C p.Q389H Chr.10: 97667099-97667099 on Build GRCh38
7141664 c.1144G>T p.D382Y Chr.10: 97667076-97667076 on Build GRCh38
4714220 c.1233C>T p.F411F Chr.10: 97667075-97667075 on Build GRCh38
4714221 c.1143C>T p.F381F Chr.10: 97667075-97667075 on Build GRCh38
6745743 c.1234G>A p.D412N Chr.10: 97667076-97667076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761522677 Chr.10: 97667080-97667080 on Build GRCh38
rs79179769 Chr.10: 97667153-97667153 on Build GRCh38
rs368876637 Chr.10: 97667139-97667139 on Build GRCh38
rs12253175 Chr.10: 97666994-97666994 on Build GRCh38
rs1002519062 Chr.10: 97666995-97666995 on Build GRCh38
rs745973463 Chr.10: 97667158-97667158 on Build GRCh38
rs746799620 Chr.10: 97667050-97667050 on Build GRCh38
rs368596585 Chr.10: 97667075-97667075 on Build GRCh38
rs779206032 Chr.10: 97667154-97667154 on Build GRCh38
rs751665193 Chr.10: 97667156-97667157 on Build GRCh38
rs1006382012 Chr.10: 97667052-97667052 on Build GRCh38
rs994595234 Chr.10: 97667169-97667169 on Build GRCh38
rs565205732 Chr.10: 97667034-97667034 on Build GRCh38
rs768473270 Chr.10: 97667063-97667063 on Build GRCh38
rs753181578 Chr.10: 97667113-97667113 on Build GRCh38
rs772713485 Chr.10: 97667101-97667101 on Build GRCh38
rs745881267 Chr.10: 97667030-97667030 on Build GRCh38
rs764457846 Chr.10: 97667054-97667054 on Build GRCh38
rs772137213 Chr.10: 97667040-97667040 on Build GRCh38
rs758500705 Chr.10: 97667162-97667162 on Build GRCh38
rs372731138 Chr.10: 97667035-97667035 on Build GRCh38
rs754068644 Chr.10: 97667144-97667144 on Build GRCh38
rs898874799 Chr.10: 97667168-97667168 on Build GRCh38
rs756478199 Chr.10: 97667117-97667117 on Build GRCh38
rs553154389 Chr.10: 97667100-97667100 on Build GRCh38
rs566581799 Chr.10: 97667049-97667049 on Build GRCh38
rs764712366 Chr.10: 97667087-97667087 on Build GRCh38
rs751856414 Chr.10: 97667112-97667112 on Build GRCh38
rs779126537 Chr.10: 97667016-97667016 on Build GRCh38
rs376952435 Chr.10: 97667172-97667172 on Build GRCh38

View Full Product Details