Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50293105-50293378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYAL3
Entrez Gene ID
8372
Gene Name
hyaluronoglucosaminidase 3
Gene Aliases
HYAL-3, LUCA-3, LUCA3
UniGene
Hs.129910
Gene Chromosome Location
Chr.3: 50292828-50299468 on Build GRCh38
Transcripts
NM_001200029.1 NM_001200030.1 NM_001200031.1 NM_001200032.1 NM_003549.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6683297 c.1138C>T p.H380Y Chr.3: 50293362-50293362 on Build GRCh38
4967374 c.1231C>G p.P411A Chr.3: 50293269-50293269 on Build GRCh38
6683295 c.1231C>T p.P411S Chr.3: 50293269-50293269 on Build GRCh38
731207 c.1222G>C p.E408Q Chr.3: 50293278-50293278 on Build GRCh38
6683294 c.1129G>A p.A377T Chr.3: 50293371-50293371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782519738 Chr.3: 50293152-50293152 on Build GRCh38
rs781944421 Chr.3: 50293200-50293200 on Build GRCh38
rs781968056 Chr.3: 50293305-50293305 on Build GRCh38
rs782393507 Chr.3: 50293127-50293127 on Build GRCh38
rs782638162 Chr.3: 50293283-50293283 on Build GRCh38
rs1029241115 Chr.3: 50293289-50293289 on Build GRCh38
rs782339965 Chr.3: 50293302-50293302 on Build GRCh38
rs891564173 Chr.3: 50293149-50293149 on Build GRCh38
rs587633640 Chr.3: 50293153-50293153 on Build GRCh38
rs782115677 Chr.3: 50293327-50293327 on Build GRCh38
rs782169093 Chr.3: 50293355-50293355 on Build GRCh38
rs935870911 Chr.3: 50293144-50293144 on Build GRCh38
rs782437665 Chr.3: 50293202-50293202 on Build GRCh38
rs782439743 Chr.3: 50293330-50293330 on Build GRCh38
rs1006828169 Chr.3: 50293348-50293348 on Build GRCh38
rs782598411 Chr.3: 50293167-50293168 on Build GRCh38
rs140479295 Chr.3: 50293290-50293290 on Build GRCh38
rs782022468 Chr.3: 50293352-50293352 on Build GRCh38
rs782595667 Chr.3: 50293276-50293277 on Build GRCh38
rs782693162 Chr.3: 50293140-50293140 on Build GRCh38
rs782435087 Chr.3: 50293271-50293272 on Build GRCh38
rs917900632 Chr.3: 50293136-50293136 on Build GRCh38
rs587758515 Chr.3: 50293151-50293151 on Build GRCh38
rs782193328 Chr.3: 50293296-50293296 on Build GRCh38
rs782682787 Chr.3: 50293188-50293188 on Build GRCh38
rs142423146 Chr.3: 50293300-50293300 on Build GRCh38
rs782519762 Chr.3: 50293126-50293126 on Build GRCh38
rs782509137 Chr.3: 50293263-50293263 on Build GRCh38
rs376595840 Chr.3: 50293252-50293252 on Build GRCh38
rs782588978 Chr.3: 50293213-50293213 on Build GRCh38
rs781805308 Chr.3: 50293142-50293142 on Build GRCh38
rs782218314 Chr.3: 50293255-50293255 on Build GRCh38
rs587694855 Chr.3: 50293333-50293333 on Build GRCh38
rs782051251 Chr.3: 50293353-50293353 on Build GRCh38
rs587680727 Chr.3: 50293137-50293137 on Build GRCh38
rs782014643 Chr.3: 50293132-50293132 on Build GRCh38
rs992590891 Chr.3: 50293133-50293133 on Build GRCh38
rs782798046 Chr.3: 50293347-50293347 on Build GRCh38

View Full Product Details