Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141384706-141384967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGA6
Entrez Gene ID
56109
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
PCDH-GAMMA-A6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141374084-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGA7
Entrez Gene ID
56108
Gene Name
protocadherin gamma subfamily A, 7
Gene Aliases
PCDH-GAMMA-A7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141382739-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGB3
Entrez Gene ID
56102
Gene Name
protocadherin gamma subfamily B, 3
Gene Aliases
PCDH-GAMMA-B3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141370264-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4473997 c.1888C>T p.R630W Chr.5: 141384787-141384787 on Build GRCh38
3851915 c.1807T>G p.W603G Chr.5: 141384706-141384706 on Build GRCh38
7155743 c.1957C>T p.P653S Chr.5: 141384856-141384856 on Build GRCh38
5444483 c.1871G>A p.G624D Chr.5: 141384770-141384770 on Build GRCh38
4473020 c.1821C>T p.L607L Chr.5: 141384720-141384720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369146747 Chr.5: 141384758-141384758 on Build GRCh38
rs750050142 Chr.5: 141384826-141384826 on Build GRCh38
rs769507654 Chr.5: 141384942-141384942 on Build GRCh38
rs768506502 Chr.5: 141384783-141384783 on Build GRCh38
rs974144646 Chr.5: 141384752-141384752 on Build GRCh38
rs773450358 Chr.5: 141384791-141384791 on Build GRCh38
rs775593747 Chr.5: 141384773-141384773 on Build GRCh38
rs769607351 Chr.5: 141384871-141384871 on Build GRCh38
rs951463054 Chr.5: 141384882-141384882 on Build GRCh38
rs750109041 Chr.5: 141384934-141384934 on Build GRCh38
rs865860988 Chr.5: 141384919-141384919 on Build GRCh38
rs747704737 Chr.5: 141384762-141384762 on Build GRCh38
rs568102289 Chr.5: 141384771-141384771 on Build GRCh38
rs766396351 Chr.5: 141384797-141384797 on Build GRCh38
rs202180798 Chr.5: 141384864-141384864 on Build GRCh38
rs781038163 Chr.5: 141384940-141384940 on Build GRCh38
rs941480085 Chr.5: 141384750-141384750 on Build GRCh38
rs149266522 Chr.5: 141384822-141384822 on Build GRCh38
rs182404532 Chr.5: 141384827-141384827 on Build GRCh38
rs982769749 Chr.5: 141384898-141384898 on Build GRCh38
rs778564034 Chr.5: 141384739-141384739 on Build GRCh38
rs866832912 Chr.5: 141384845-141384845 on Build GRCh38
rs746957314 Chr.5: 141384828-141384828 on Build GRCh38
rs773444317 Chr.5: 141384872-141384872 on Build GRCh38
rs753215361 Chr.5: 141384907-141384907 on Build GRCh38
rs868472813 Chr.5: 141384908-141384908 on Build GRCh38
rs771003299 Chr.5: 141384879-141384879 on Build GRCh38
rs781370362 Chr.5: 141384749-141384749 on Build GRCh38
rs766909881 Chr.5: 141384727-141384727 on Build GRCh38
rs755407612 Chr.5: 141384810-141384810 on Build GRCh38
rs748067928 Chr.5: 141384941-141384941 on Build GRCh38
rs1027171096 Chr.5: 141384853-141384853 on Build GRCh38
rs764481830 Chr.5: 141384930-141384930 on Build GRCh38
rs776231525 Chr.5: 141384785-141384785 on Build GRCh38
rs570988671 Chr.5: 141384825-141384825 on Build GRCh38
rs373370095 Chr.5: 141384817-141384817 on Build GRCh38
rs755356826 Chr.5: 141384747-141384747 on Build GRCh38
rs369584315 Chr.5: 141384834-141384834 on Build GRCh38
rs374905153 Chr.5: 141384792-141384792 on Build GRCh38
rs761655907 Chr.5: 141384787-141384787 on Build GRCh38
rs752038561 Chr.5: 141384730-141384730 on Build GRCh38
rs760972840 Chr.5: 141384922-141384922 on Build GRCh38
rs373048330 Chr.5: 141384937-141384937 on Build GRCh38
rs191053275 Chr.5: 141384800-141384800 on Build GRCh38
rs999175494 Chr.5: 141384790-141384790 on Build GRCh38
rs959887704 Chr.5: 141384794-141384794 on Build GRCh38
rs777478209 Chr.5: 141384943-141384943 on Build GRCh38
rs1052082057 Chr.5: 141384801-141384801 on Build GRCh38
rs759644846 Chr.5: 141384884-141384884 on Build GRCh38
rs371823901 Chr.5: 141384767-141384767 on Build GRCh38
rs774353364 Chr.5: 141384883-141384883 on Build GRCh38
rs778610936 Chr.5: 141384769-141384769 on Build GRCh38
rs746906389 Chr.5: 141384782-141384782 on Build GRCh38
rs781186343 Chr.5: 141384847-141384847 on Build GRCh38
rs767972748 Chr.5: 141384906-141384906 on Build GRCh38
rs1040265836 Chr.5: 141384748-141384749 on Build GRCh38
rs763067876 Chr.5: 141384795-141384795 on Build GRCh38
rs765446040 Chr.5: 141384788-141384788 on Build GRCh38
rs114533608 Chr.5: 141384753-141384753 on Build GRCh38
rs573213865 Chr.5: 141384870-141384870 on Build GRCh38
rs754481008 Chr.5: 141384938-141384938 on Build GRCh38
rs1046016115 Chr.5: 141384768-141384768 on Build GRCh38

View Full Product Details