Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 128040787-128041054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRE
Entrez Gene ID
5791
Gene Name
protein tyrosine phosphatase, receptor type E
Gene Aliases
HPTPE, PTPE, R-PTP-EPSILON
UniGene
Hs.127022
Gene Chromosome Location
Chr.10: 127907061-128085855 on Build GRCh38
Transcripts
NM_001316676.1 NM_001316677.1 NM_001323354.1 NM_001323355.1 NM_001323356.1 NM_006504.5 XM_005252691.2 XM_011539994.2 XM_011539995.1 XM_011539996.1 XM_011539998.1 XM_017016467.1 XM_017016468.1 XM_017016469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
142811 c.32G>A p.G11D Chr.10: 128040913-128040913 on Build GRCh38
6893193 c.51C>T p.A17A Chr.10: 128040932-128040932 on Build GRCh38
1346736 c.53G>T p.R18M Chr.10: 128040934-128040934 on Build GRCh38
5561680 c.44C>T p.P15L Chr.10: 128040925-128040925 on Build GRCh38
7164471 c.21C>T p.L7L Chr.10: 128040902-128040902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745907322 Chr.10: 128040872-128040872 on Build GRCh38
rs759075580 Chr.10: 128040983-128040983 on Build GRCh38
rs751504741 Chr.10: 128041000-128041000 on Build GRCh38
rs568464160 Chr.10: 128040817-128040817 on Build GRCh38
rs752301484 Chr.10: 128040997-128040997 on Build GRCh38
rs780342381 Chr.10: 128040962-128040962 on Build GRCh38
rs377165003 Chr.10: 128040963-128040963 on Build GRCh38
rs201481452 Chr.10: 128040845-128040845 on Build GRCh38
rs764817797 Chr.10: 128040984-128040984 on Build GRCh38
rs758147050 Chr.10: 128040940-128040940 on Build GRCh38
rs1045154634 Chr.10: 128041024-128041024 on Build GRCh38
rs760163795 Chr.10: 128040914-128040914 on Build GRCh38
rs371474905 Chr.10: 128040842-128040842 on Build GRCh38
rs751460267 Chr.10: 128040950-128040950 on Build GRCh38
rs748011312 Chr.10: 128040856-128040856 on Build GRCh38
rs370004056 Chr.10: 128040982-128040982 on Build GRCh38
rs374843074 Chr.10: 128040893-128040893 on Build GRCh38
rs898467409 Chr.10: 128040883-128040883 on Build GRCh38
rs949488179 Chr.10: 128041020-128041020 on Build GRCh38
rs535424361 Chr.10: 128040818-128040818 on Build GRCh38
rs75709098 Chr.10: 128040929-128040929 on Build GRCh38
rs576132497 Chr.10: 128040951-128040951 on Build GRCh38
rs761143024 Chr.10: 128040980-128040980 on Build GRCh38
rs773764698 Chr.10: 128040974-128040974 on Build GRCh38
rs757316163 Chr.10: 128041002-128041002 on Build GRCh38
rs753930772 Chr.10: 128041019-128041019 on Build GRCh38
rs766915661 Chr.10: 128040830-128040830 on Build GRCh38
rs144835853 Chr.10: 128040971-128040971 on Build GRCh38
rs769779567 Chr.10: 128040810-128040810 on Build GRCh38
rs557975239 Chr.10: 128040945-128040945 on Build GRCh38
rs765939778 Chr.10: 128040919-128040919 on Build GRCh38
rs774906552 Chr.10: 128040902-128040902 on Build GRCh38
rs371825910 Chr.10: 128040955-128040955 on Build GRCh38
rs762196023 Chr.10: 128040911-128040911 on Build GRCh38
rs749991050 Chr.10: 128040832-128040832 on Build GRCh38
rs777599354 Chr.10: 128040871-128040871 on Build GRCh38
rs762746774 Chr.10: 128040999-128040999 on Build GRCh38
rs772328329 Chr.10: 128040972-128040972 on Build GRCh38
rs753474086 Chr.10: 128040926-128040926 on Build GRCh38
rs953038425 Chr.10: 128040922-128040922 on Build GRCh38
rs971931971 Chr.10: 128040979-128040979 on Build GRCh38
rs199705745 Chr.10: 128040934-128040934 on Build GRCh38
rs865899773 Chr.10: 128040961-128040961 on Build GRCh38
rs921629125 Chr.10: 128040907-128040907 on Build GRCh38
rs768029326 Chr.10: 128040912-128040912 on Build GRCh38
rs1020159531 Chr.10: 128040841-128040841 on Build GRCh38
rs764982543 Chr.10: 128040930-128040930 on Build GRCh38
rs753492579 Chr.10: 128040846-128040846 on Build GRCh38
rs1025685992 Chr.10: 128040928-128040928 on Build GRCh38
rs267602412 Chr.10: 128040913-128040913 on Build GRCh38
rs758397687 Chr.10: 128040863-128040863 on Build GRCh38
rs977472597 Chr.10: 128040971-128040971 on Build GRCh38
rs963846338 Chr.10: 128040954-128040954 on Build GRCh38
rs780252405 Chr.10: 128041003-128041003 on Build GRCh38
rs947061250 Chr.10: 128040809-128040809 on Build GRCh38
rs775791967 Chr.10: 128040879-128040879 on Build GRCh38
rs370425175 Chr.10: 128040852-128040852 on Build GRCh38
rs773454404 Chr.10: 128040814-128040814 on Build GRCh38
rs769780836 Chr.10: 128040875-128040875 on Build GRCh38

View Full Product Details