Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220153066-220153295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB3GAP2
Entrez Gene ID
25782
Gene Name
RAB3 GTPase activating non-catalytic protein subunit 2
Gene Aliases
RAB3-GAP150, RAB3GAP150, SPG69, WARBM2, p150
UniGene
Hs.654849
Gene Chromosome Location
Chr.1: 220148268-220272501 on Build GRCh38
Transcripts
NM_012414.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3803947 c.3859G>A p.G1287R Chr.1: 220153193-220153193 on Build GRCh38
3705532 c.3832C>T p.L1278L Chr.1: 220153220-220153220 on Build GRCh38
1738531 c.3761C>T p.A1254V Chr.1: 220153291-220153291 on Build GRCh38
6378062 c.3847G>A p.V1283I Chr.1: 220153205-220153205 on Build GRCh38
4633580 c.3766G>T p.A1256S Chr.1: 220153286-220153286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776392076 Chr.1: 220153159-220153159 on Build GRCh38
rs768043319 Chr.1: 220153175-220153175 on Build GRCh38
rs374980242 Chr.1: 220153145-220153145 on Build GRCh38
rs201231700 Chr.1: 220153146-220153146 on Build GRCh38
rs886046019 Chr.1: 220153223-220153223 on Build GRCh38
rs746994578 Chr.1: 220153149-220153149 on Build GRCh38
rs762682611 Chr.1: 220153174-220153174 on Build GRCh38
rs370057302 Chr.1: 220153234-220153234 on Build GRCh38
rs748424129 Chr.1: 220153248-220153248 on Build GRCh38
rs543919777 Chr.1: 220153271-220153271 on Build GRCh38
rs200579008 Chr.1: 220153172-220153172 on Build GRCh38
rs985161879 Chr.1: 220153189-220153189 on Build GRCh38
rs371927934 Chr.1: 220153165-220153165 on Build GRCh38
rs34081806 Chr.1: 220153210-220153210 on Build GRCh38
rs748299419 Chr.1: 220153135-220153135 on Build GRCh38
rs778851334 Chr.1: 220153247-220153247 on Build GRCh38
rs114940727 Chr.1: 220153152-220153152 on Build GRCh38
rs940729257 Chr.1: 220153227-220153227 on Build GRCh38
rs766935556 Chr.1: 220153222-220153222 on Build GRCh38
rs373515509 Chr.1: 220153255-220153255 on Build GRCh38
rs771919095 Chr.1: 220153140-220153140 on Build GRCh38
rs764592566 Chr.1: 220153170-220153170 on Build GRCh38
rs756510210 Chr.1: 220153217-220153217 on Build GRCh38
rs866811119 Chr.1: 220153273-220153273 on Build GRCh38
rs921840551 Chr.1: 220153156-220153156 on Build GRCh38
rs755092641 Chr.1: 220153246-220153246 on Build GRCh38

View Full Product Details