Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48446434-48446707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRWD1
Entrez Gene ID
83743
Gene Name
glutamate rich WD repeat containing 1
Gene Aliases
CDW4, GRWD, RRB1, WDR28
UniGene
Hs.400625
Gene Chromosome Location
Chr.19: 48445773-48453907 on Build GRCh38
Transcripts
NM_031485.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6676322 c.293C>T p.A98V Chr.19: 48446490-48446490 on Build GRCh38
5822184 c.272C>G p.A91G Chr.19: 48446469-48446469 on Build GRCh38
3107358 c.296A>C p.Q99P Chr.19: 48446493-48446493 on Build GRCh38
148681 c.305+7C>G p.? Chr.19: 48446509-48446509 on Build GRCh38
4980767 c.238C>T p.R80W Chr.19: 48446435-48446435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201051910 Chr.19: 48446551-48446551 on Build GRCh38
rs140240394 Chr.19: 48446488-48446488 on Build GRCh38
rs941200716 Chr.19: 48446682-48446682 on Build GRCh38
rs961077962 Chr.19: 48446487-48446487 on Build GRCh38
rs755737651 Chr.19: 48446655-48446655 on Build GRCh38
rs775223033 Chr.19: 48446471-48446471 on Build GRCh38
rs368404768 Chr.19: 48446631-48446631 on Build GRCh38
rs371836169 Chr.19: 48446678-48446678 on Build GRCh38
rs1035850861 Chr.19: 48446459-48446459 on Build GRCh38
rs3760795 Chr.19: 48446507-48446507 on Build GRCh38
rs777390325 Chr.19: 48446656-48446656 on Build GRCh38
rs370278405 Chr.19: 48446538-48446538 on Build GRCh38
rs774235436 Chr.19: 48446458-48446458 on Build GRCh38
rs759203855 Chr.19: 48446464-48446464 on Build GRCh38
rs748768419 Chr.19: 48446663-48446663 on Build GRCh38
rs923319292 Chr.19: 48446617-48446617 on Build GRCh38
rs1002664727 Chr.19: 48446456-48446456 on Build GRCh38
rs73588089 Chr.19: 48446591-48446591 on Build GRCh38
rs1017877855 Chr.19: 48446592-48446592 on Build GRCh38
rs950856457 Chr.19: 48446634-48446634 on Build GRCh38
rs964668269 Chr.19: 48446600-48446600 on Build GRCh38
rs778372921 Chr.19: 48446685-48446685 on Build GRCh38
rs976096953 Chr.19: 48446610-48446610 on Build GRCh38
rs763152638 Chr.19: 48446648-48446648 on Build GRCh38
rs929743 Chr.19: 48446509-48446509 on Build GRCh38
rs115286916 Chr.19: 48446535-48446535 on Build GRCh38

View Full Product Details