Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10317249-10317496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAVER1
Entrez Gene ID
125950
Gene Name
ribonucleoprotein, PTB binding 1
Gene Aliases
-
UniGene
Hs.744952
Gene Chromosome Location
Chr.19: 10316212-10333638 on Build GRCh38
Transcripts
NM_133452.2 XM_011527671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6763419 c.2185G>A p.A729T Chr.19: 10317489-10317489 on Build GRCh38
323000 c.2180A>G p.H727R Chr.19: 10317494-10317494 on Build GRCh38
1589617 c.2238G>A p.A746A Chr.19: 10317487-10317487 on Build GRCh38
1193287 c.2239G>C p.D747H Chr.19: 10317486-10317486 on Build GRCh38
6763418 c.2236G>A p.A746T Chr.19: 10317489-10317489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932162869 Chr.19: 10317373-10317374 on Build GRCh38
rs969220731 Chr.19: 10317440-10317440 on Build GRCh38
rs890545670 Chr.19: 10317377-10317377 on Build GRCh38
rs1024044465 Chr.19: 10317350-10317350 on Build GRCh38
rs763456832 Chr.19: 10317405-10317405 on Build GRCh38
rs767510405 Chr.19: 10317434-10317434 on Build GRCh38
rs759672865 Chr.19: 10317422-10317422 on Build GRCh38
rs981860871 Chr.19: 10317311-10317311 on Build GRCh38
rs914951672 Chr.19: 10317346-10317346 on Build GRCh38
rs1014526237 Chr.19: 10317298-10317298 on Build GRCh38
rs771528534 Chr.19: 10317413-10317413 on Build GRCh38
rs769804911 Chr.19: 10317354-10317354 on Build GRCh38
rs988216707 Chr.19: 10317337-10317345 on Build GRCh38
rs1023410024 Chr.19: 10317279-10317279 on Build GRCh38
rs961254199 Chr.19: 10317292-10317292 on Build GRCh38
rs1005005494 Chr.19: 10317404-10317404 on Build GRCh38
rs200027669 Chr.19: 10317471-10317471 on Build GRCh38
rs566583285 Chr.19: 10317374-10317374 on Build GRCh38
rs1015406482 Chr.19: 10317286-10317286 on Build GRCh38
rs927715130 Chr.19: 10317312-10317312 on Build GRCh38
rs867065326 Chr.19: 10317458-10317458 on Build GRCh38
rs760842191 Chr.19: 10317443-10317443 on Build GRCh38
rs922079313 Chr.19: 10317370-10317370 on Build GRCh38
rs748597660 Chr.19: 10317390-10317391 on Build GRCh38
rs368364557 Chr.19: 10317416-10317416 on Build GRCh38
rs1043358947 Chr.19: 10317369-10317369 on Build GRCh38
rs958246747 Chr.19: 10317435-10317435 on Build GRCh38
rs752900668 Chr.19: 10317437-10317437 on Build GRCh38
rs756601273 Chr.19: 10317404-10317405 on Build GRCh38
rs925622090 Chr.19: 10317383-10317383 on Build GRCh38
rs763902015 Chr.19: 10317453-10317453 on Build GRCh38
rs956364841 Chr.19: 10317330-10317330 on Build GRCh38

View Full Product Details