Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23196541-23196812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUPL2
Entrez Gene ID
11097
Gene Name
nucleoporin like 2
Gene Aliases
CG1, NLP-1, NLP_1, hCG1
UniGene
Hs.408241
Gene Chromosome Location
Chr.7: 23181567-23201011 on Build GRCh38
Transcripts
NM_007342.2 XM_005249592.1 XM_005249593.3 XM_011515104.2 XM_017011714.1 XM_017011715.1 XM_017011716.1 XM_017011717.1 XM_017011718.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6731776 c.572A>C p.E191A Chr.7: 23196729-23196729 on Build GRCh38
3637509 c.530C>T p.S177F Chr.7: 23196687-23196687 on Build GRCh38
5676284 c.579A>T p.K193N Chr.7: 23196736-23196736 on Build GRCh38
1088580 c.530C>A p.S177Y Chr.7: 23196687-23196687 on Build GRCh38
1088581 c.539G>A p.R180H Chr.7: 23196696-23196696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879140978 Chr.7: 23196780-23196780 on Build GRCh38
rs1009430227 Chr.7: 23196599-23196599 on Build GRCh38
rs763615256 Chr.7: 23196696-23196696 on Build GRCh38
rs755435290 Chr.7: 23196747-23196747 on Build GRCh38
rs552017909 Chr.7: 23196748-23196748 on Build GRCh38
rs778001251 Chr.7: 23196774-23196774 on Build GRCh38
rs751114640 Chr.7: 23196705-23196705 on Build GRCh38
rs957494079 Chr.7: 23196737-23196737 on Build GRCh38
rs376419470 Chr.7: 23196629-23196629 on Build GRCh38
rs771166226 Chr.7: 23196784-23196784 on Build GRCh38
rs752567714 Chr.7: 23196596-23196596 on Build GRCh38
rs770562467 Chr.7: 23196665-23196665 on Build GRCh38
rs186355447 Chr.7: 23196745-23196745 on Build GRCh38
rs3735238 Chr.7: 23196663-23196663 on Build GRCh38
rs549961021 Chr.7: 23196574-23196574 on Build GRCh38
rs76452127 Chr.7: 23196783-23196783 on Build GRCh38
rs201232262 Chr.7: 23196671-23196671 on Build GRCh38
rs374167066 Chr.7: 23196678-23196678 on Build GRCh38
rs753281820 Chr.7: 23196755-23196755 on Build GRCh38
rs759303991 Chr.7: 23196672-23196672 on Build GRCh38
rs745831723 Chr.7: 23196744-23196745 on Build GRCh38
rs761524771 Chr.7: 23196710-23196710 on Build GRCh38
rs75513762 Chr.7: 23196728-23196728 on Build GRCh38
rs966066540 Chr.7: 23196597-23196597 on Build GRCh38
rs535306272 Chr.7: 23196687-23196687 on Build GRCh38
rs767136071 Chr.7: 23196712-23196712 on Build GRCh38
rs372545705 Chr.7: 23196769-23196769 on Build GRCh38
rs368253769 Chr.7: 23196702-23196702 on Build GRCh38
rs759066974 Chr.7: 23196600-23196600 on Build GRCh38
rs769693804 Chr.7: 23196689-23196689 on Build GRCh38
rs775445241 Chr.7: 23196694-23196694 on Build GRCh38
rs762409900 Chr.7: 23196695-23196695 on Build GRCh38
rs1020904166 Chr.7: 23196746-23196746 on Build GRCh38
rs769140088 Chr.7: 23196782-23196783 on Build GRCh38
rs1048237572 Chr.7: 23196581-23196581 on Build GRCh38

View Full Product Details