Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 98058-98331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPRL3
Entrez Gene ID
8131
Gene Name
NPR3 like, GATOR1 complex subunit
Gene Aliases
C16orf35, CGTHBA, HS-40, MARE, NPR3, RMD11
UniGene
Hs.19699
Gene Chromosome Location
Chr.16: 85801-138698 on Build GRCh38
Transcripts
NM_001039476.2 NM_001077350.2 NM_001243247.1 NM_001243248.1 NM_001243249.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6728888 c.851G>A p.R284Q Chr.16: 98218-98218 on Build GRCh38
283348 c.801G>T p.E267D Chr.16: 98268-98268 on Build GRCh38
2144018 c.850C>T p.R284W Chr.16: 98219-98219 on Build GRCh38
967263 c.824C>A p.P275H Chr.16: 98245-98245 on Build GRCh38
7258127 c.924G>T p.Q308H Chr.16: 98145-98145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756427551 Chr.16: 98128-98128 on Build GRCh38
rs746568841 Chr.16: 98300-98300 on Build GRCh38
rs888753529 Chr.16: 98081-98081 on Build GRCh38
rs777449086 Chr.16: 98272-98272 on Build GRCh38
rs757942313 Chr.16: 98250-98250 on Build GRCh38
rs569165571 Chr.16: 98104-98104 on Build GRCh38
rs768908442 Chr.16: 98200-98200 on Build GRCh38
rs777790535 Chr.16: 98164-98164 on Build GRCh38
rs767954646 Chr.16: 98109-98109 on Build GRCh38
rs867730506 Chr.16: 98295-98295 on Build GRCh38
rs566886246 Chr.16: 98311-98311 on Build GRCh38
rs745692079 Chr.16: 98193-98193 on Build GRCh38
rs764273616 Chr.16: 98099-98099 on Build GRCh38
rs754229199 Chr.16: 98223-98223 on Build GRCh38
rs1026188399 Chr.16: 98141-98141 on Build GRCh38
rs755359263 Chr.16: 98225-98225 on Build GRCh38
rs375410124 Chr.16: 98254-98254 on Build GRCh38
rs752301363 Chr.16: 98249-98249 on Build GRCh38
rs780415433 Chr.16: 98129-98129 on Build GRCh38
rs764890667 Chr.16: 98243-98243 on Build GRCh38
rs984688777 Chr.16: 98258-98258 on Build GRCh38
rs767611082 Chr.16: 98208-98208 on Build GRCh38
rs774447369 Chr.16: 98206-98206 on Build GRCh38
rs11558706 Chr.16: 98240-98240 on Build GRCh38
rs754657607 Chr.16: 98131-98131 on Build GRCh38
rs771726316 Chr.16: 98163-98163 on Build GRCh38
rs549291288 Chr.16: 98150-98150 on Build GRCh38
rs770800767 Chr.16: 98175-98175 on Build GRCh38
rs776547727 Chr.16: 98188-98188 on Build GRCh38
rs550763256 Chr.16: 98098-98098 on Build GRCh38
rs866111108 Chr.16: 98080-98080 on Build GRCh38
rs886037958 Chr.16: 98233-98234 on Build GRCh38
rs750933514 Chr.16: 98115-98115 on Build GRCh38
rs778603279 Chr.16: 98226-98226 on Build GRCh38
rs746975723 Chr.16: 98171-98171 on Build GRCh38
rs754625313 Chr.16: 98134-98134 on Build GRCh38
rs766908016 Chr.16: 98219-98219 on Build GRCh38
rs757231242 Chr.16: 98304-98304 on Build GRCh38
rs373936494 Chr.16: 98216-98216 on Build GRCh38
rs369959139 Chr.16: 98213-98213 on Build GRCh38
rs761854585 Chr.16: 98108-98108 on Build GRCh38
rs778499898 Chr.16: 98138-98138 on Build GRCh38
rs1033607990 Chr.16: 98262-98262 on Build GRCh38
rs773656881 Chr.16: 98209-98209 on Build GRCh38

View Full Product Details