Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76570469-76570742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQGAP2
Entrez Gene ID
10788
Gene Name
IQ motif containing GTPase activating protein 2
Gene Aliases
-
UniGene
Hs.291030
Gene Chromosome Location
Chr.5: 76403255-76708132 on Build GRCh38
Transcripts
NM_001285460.1 NM_006633.3 XM_005248410.2 XM_017008960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4586006 c.340G>A p.V114I Chr.5: 76570616-76570616 on Build GRCh38
5996280 c.381+10C>T p.? Chr.5: 76570667-76570667 on Build GRCh38
78969 c.343C>T p.Q115* Chr.5: 76570619-76570619 on Build GRCh38
1567604 c.304-1G>C p.? Chr.5: 76570579-76570579 on Build GRCh38
461950 c.342C>G p.V114V Chr.5: 76570618-76570618 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200990534 Chr.5: 76570658-76570658 on Build GRCh38
rs778273441 Chr.5: 76570616-76570616 on Build GRCh38
rs370471380 Chr.5: 76570555-76570555 on Build GRCh38
rs115631027 Chr.5: 76570615-76570615 on Build GRCh38
rs779932610 Chr.5: 76570540-76570540 on Build GRCh38
rs375571652 Chr.5: 76570693-76570693 on Build GRCh38
rs773155176 Chr.5: 76570648-76570648 on Build GRCh38
rs192694811 Chr.5: 76570598-76570598 on Build GRCh38
rs764865648 Chr.5: 76570599-76570599 on Build GRCh38
rs746302129 Chr.5: 76570633-76570633 on Build GRCh38
rs757563873 Chr.5: 76570627-76570627 on Build GRCh38
rs752236444 Chr.5: 76570612-76570612 on Build GRCh38
rs377105589 Chr.5: 76570554-76570554 on Build GRCh38
rs769815010 Chr.5: 76570503-76570503 on Build GRCh38
rs1029248183 Chr.5: 76570687-76570687 on Build GRCh38
rs770047478 Chr.5: 76570548-76570548 on Build GRCh38
rs768651255 Chr.5: 76570642-76570642 on Build GRCh38
rs375424418 Chr.5: 76570569-76570569 on Build GRCh38
rs775920127 Chr.5: 76570541-76570542 on Build GRCh38
rs780155939 Chr.5: 76570638-76570638 on Build GRCh38
rs1038302660 Chr.5: 76570647-76570647 on Build GRCh38
rs991181510 Chr.5: 76570524-76570524 on Build GRCh38
rs146065067 Chr.5: 76570632-76570632 on Build GRCh38
rs749325481 Chr.5: 76570640-76570640 on Build GRCh38
rs753736233 Chr.5: 76570589-76570589 on Build GRCh38
rs776791619 Chr.5: 76570537-76570537 on Build GRCh38
rs372285513 Chr.5: 76570669-76570669 on Build GRCh38
rs748824629 Chr.5: 76570541-76570541 on Build GRCh38
rs766299341 Chr.5: 76570583-76570583 on Build GRCh38
rs201969391 Chr.5: 76570668-76570668 on Build GRCh38
rs767434046 Chr.5: 76570567-76570567 on Build GRCh38
rs368635855 Chr.5: 76570700-76570700 on Build GRCh38
rs968090876 Chr.5: 76570709-76570709 on Build GRCh38
rs544252052 Chr.5: 76570618-76570618 on Build GRCh38
rs751115627 Chr.5: 76570707-76570707 on Build GRCh38
rs80247589 Chr.5: 76570670-76570670 on Build GRCh38
rs989422308 Chr.5: 76570593-76570593 on Build GRCh38
rs943987553 Chr.5: 76570660-76570660 on Build GRCh38
rs367622453 Chr.5: 76570570-76570570 on Build GRCh38

View Full Product Details