Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 157442544-157442809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTX3
Entrez Gene ID
5806
Gene Name
pentraxin 3
Gene Aliases
TNFAIP5, TSG-14
UniGene
Hs.591286
Gene Chromosome Location
Chr.3: 157436791-157443628 on Build GRCh38
Transcripts
NM_002852.3 NM_001167911.1 NM_001167912.1 NM_024621.2 XM_011513134.2 XM_011513135.1

Gene Symbol
VEPH1
Entrez Gene ID
79674
Gene Name
ventricular zone expressed PH domain containing 1
Gene Aliases
MELT, VEPH
UniGene
Hs.658046
Gene Chromosome Location
Chr.3: 157259742-157503626 on Build GRCh38
Transcripts
NM_002852.3 NM_001167911.1 NM_001167912.1 NM_024621.2 XM_011513134.2 XM_011513135.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
729489 c.859G>C p.G287R Chr.3: 157442692-157442692 on Build GRCh38
1180325 c.870G>T p.A290A Chr.3: 157442703-157442703 on Build GRCh38
5742528 c.869C>T p.A290V Chr.3: 157442702-157442702 on Build GRCh38
4562233 c.917G>A p.G306E Chr.3: 157442750-157442750 on Build GRCh38
6468594 c.942G>A p.K314K Chr.3: 157442775-157442775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143287219 Chr.3: 157442752-157442752 on Build GRCh38
rs373914748 Chr.3: 157442781-157442781 on Build GRCh38
rs934868526 Chr.3: 157442713-157442713 on Build GRCh38
rs767120397 Chr.3: 157442675-157442675 on Build GRCh38
rs919038712 Chr.3: 157442639-157442639 on Build GRCh38
rs951755578 Chr.3: 157442677-157442677 on Build GRCh38
rs745911276 Chr.3: 157442749-157442749 on Build GRCh38
rs769709134 Chr.3: 157442771-157442771 on Build GRCh38
rs963132311 Chr.3: 157442573-157442573 on Build GRCh38
rs748466924 Chr.3: 157442595-157442595 on Build GRCh38
rs144979346 Chr.3: 157442750-157442750 on Build GRCh38
rs146001185 Chr.3: 157442703-157442703 on Build GRCh38
rs56265729 Chr.3: 157442569-157442569 on Build GRCh38
rs781518867 Chr.3: 157442590-157442590 on Build GRCh38
rs1052406017 Chr.3: 157442588-157442588 on Build GRCh38
rs747951421 Chr.3: 157442572-157442574 on Build GRCh38
rs987133414 Chr.3: 157442653-157442653 on Build GRCh38
rs139851251 Chr.3: 157442741-157442741 on Build GRCh38
rs774553750 Chr.3: 157442624-157442624 on Build GRCh38
rs926130606 Chr.3: 157442704-157442704 on Build GRCh38
rs757818668 Chr.3: 157442729-157442729 on Build GRCh38
rs772385247 Chr.3: 157442630-157442630 on Build GRCh38
rs778166071 Chr.3: 157442708-157442708 on Build GRCh38
rs899979858 Chr.3: 157442633-157442633 on Build GRCh38
rs4478039 Chr.3: 157442770-157442770 on Build GRCh38
rs141322370 Chr.3: 157442628-157442628 on Build GRCh38
rs190837481 Chr.3: 157442638-157442638 on Build GRCh38
rs778188616 Chr.3: 157442614-157442614 on Build GRCh38
rs370077547 Chr.3: 157442611-157442611 on Build GRCh38
rs770857279 Chr.3: 157442786-157442786 on Build GRCh38
rs773182710 Chr.3: 157442773-157442773 on Build GRCh38
rs3749295 Chr.3: 157442769-157442769 on Build GRCh38
rs35415718 Chr.3: 157442702-157442702 on Build GRCh38
rs747739786 Chr.3: 157442583-157442583 on Build GRCh38
rs879192748 Chr.3: 157442582-157442582 on Build GRCh38
rs148494144 Chr.3: 157442647-157442647 on Build GRCh38
rs144589446 Chr.3: 157442646-157442646 on Build GRCh38
rs775876046 Chr.3: 157442631-157442631 on Build GRCh38
rs754473132 Chr.3: 157442657-157442657 on Build GRCh38
rs765775634 Chr.3: 157442648-157442648 on Build GRCh38
rs765126886 Chr.3: 157442643-157442643 on Build GRCh38
rs770899550 Chr.3: 157442620-157442620 on Build GRCh38
rs752668450 Chr.3: 157442692-157442692 on Build GRCh38
rs980722173 Chr.3: 157442784-157442784 on Build GRCh38
rs749750187 Chr.3: 157442725-157442725 on Build GRCh38
rs749806282 Chr.3: 157442618-157442618 on Build GRCh38

View Full Product Details