Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 52940572-52940809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPBWR1
Entrez Gene ID
2831
Gene Name
neuropeptides B/W receptor 1
Gene Aliases
GPR7
UniGene
Hs.248117
Gene Chromosome Location
Chr.8: 52939908-52940894 on Build GRCh38
Transcripts
NM_005285.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6181297 c.669T>C p.Y223Y Chr.8: 52940576-52940576 on Build GRCh38
3413060 c.829A>G p.T277A Chr.8: 52940736-52940736 on Build GRCh38
6181296 c.798C>A p.P266P Chr.8: 52940705-52940705 on Build GRCh38
4626243 c.840G>A p.P280P Chr.8: 52940747-52940747 on Build GRCh38
4618775 c.891C>T p.Y297Y Chr.8: 52940798-52940798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959390087 Chr.8: 52940702-52940702 on Build GRCh38
rs778587874 Chr.8: 52940665-52940665 on Build GRCh38
rs757177229 Chr.8: 52940591-52940591 on Build GRCh38
rs761396667 Chr.8: 52940720-52940720 on Build GRCh38
rs369147724 Chr.8: 52940683-52940683 on Build GRCh38
rs906189485 Chr.8: 52940638-52940638 on Build GRCh38
rs773698510 Chr.8: 52940622-52940622 on Build GRCh38
rs750226588 Chr.8: 52940736-52940736 on Build GRCh38
rs373362045 Chr.8: 52940652-52940652 on Build GRCh38
rs1024540931 Chr.8: 52940603-52940603 on Build GRCh38
rs376886430 Chr.8: 52940719-52940719 on Build GRCh38
rs142975757 Chr.8: 52940696-52940696 on Build GRCh38
rs146623388 Chr.8: 52940717-52940717 on Build GRCh38
rs753629757 Chr.8: 52940656-52940656 on Build GRCh38
rs745800484 Chr.8: 52940599-52940599 on Build GRCh38
rs528274606 Chr.8: 52940621-52940621 on Build GRCh38
rs771274567 Chr.8: 52940691-52940691 on Build GRCh38
rs760053343 Chr.8: 52940710-52940710 on Build GRCh38
rs1003180345 Chr.8: 52940639-52940639 on Build GRCh38
rs774862355 Chr.8: 52940630-52940630 on Build GRCh38
rs778073538 Chr.8: 52940686-52940686 on Build GRCh38
rs369302191 Chr.8: 52940633-52940633 on Build GRCh38
rs765042857 Chr.8: 52940734-52940734 on Build GRCh38
rs756283476 Chr.8: 52940617-52940618 on Build GRCh38
rs371447815 Chr.8: 52940651-52940651 on Build GRCh38
rs758333983 Chr.8: 52940751-52940751 on Build GRCh38
rs752240874 Chr.8: 52940641-52940641 on Build GRCh38
rs201583276 Chr.8: 52940682-52940682 on Build GRCh38
rs759980764 Chr.8: 52940631-52940631 on Build GRCh38
rs768300813 Chr.8: 52940711-52940711 on Build GRCh38
rs530729259 Chr.8: 52940598-52940598 on Build GRCh38
rs757012389 Chr.8: 52940661-52940661 on Build GRCh38
rs909011928 Chr.8: 52940660-52940660 on Build GRCh38
rs749531993 Chr.8: 52940690-52940690 on Build GRCh38
rs148401487 Chr.8: 52940673-52940673 on Build GRCh38
rs75387626 Chr.8: 52940600-52940600 on Build GRCh38
rs376561808 Chr.8: 52940680-52940680 on Build GRCh38
rs774972357 Chr.8: 52940713-52940713 on Build GRCh38
rs755746368 Chr.8: 52940650-52940650 on Build GRCh38
rs756203473 Chr.8: 52940774-52940774 on Build GRCh38
rs199589648 Chr.8: 52940790-52940790 on Build GRCh38
rs748341323 Chr.8: 52940615-52940615 on Build GRCh38
rs199533493 Chr.8: 52940604-52940604 on Build GRCh38
rs541917349 Chr.8: 52940629-52940629 on Build GRCh38
rs780207150 Chr.8: 52940752-52940752 on Build GRCh38
rs770181284 Chr.8: 52940616-52940616 on Build GRCh38
rs141356626 Chr.8: 52940769-52940769 on Build GRCh38
rs1052703430 Chr.8: 52940723-52940723 on Build GRCh38
rs267601946 Chr.8: 52940756-52940756 on Build GRCh38
rs567536404 Chr.8: 52940607-52940607 on Build GRCh38

View Full Product Details