Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 123469073-123469304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQUB
Entrez Gene ID
154865
Gene Name
IQ motif and ubiquitin domain containing
Gene Aliases
-
UniGene
Hs.159650
Gene Chromosome Location
Chr.7: 123452181-123535077 on Build GRCh38
Transcripts
NM_001282855.1 NM_001321293.1 NM_178827.4 NR_104244.1 NR_104245.1 XM_005250161.3 XM_005250162.4 XM_011515833.2 XM_011515834.2 XM_017011771.1 XM_017011772.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5646934 c.1511G>A p.C504Y Chr.7: 123469284-123469284 on Build GRCh38
1210881 c.1537G>T p.D513Y Chr.7: 123469258-123469258 on Build GRCh38
5518791 c.1526delA p.N509fs*12 Chr.7: 123469269-123469269 on Build GRCh38
3877717 c.1553T>C p.V518A Chr.7: 123469242-123469242 on Build GRCh38
3632649 c.1558T>G p.L520V Chr.7: 123469237-123469237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763369359 Chr.7: 123469230-123469230 on Build GRCh38
rs776583820 Chr.7: 123469221-123469221 on Build GRCh38
rs758184950 Chr.7: 123469128-123469128 on Build GRCh38
rs771969806 Chr.7: 123469200-123469200 on Build GRCh38
rs765698354 Chr.7: 123469168-123469169 on Build GRCh38
rs911211617 Chr.7: 123469261-123469261 on Build GRCh38
rs760524191 Chr.7: 123469214-123469214 on Build GRCh38
rs10243987 Chr.7: 123469188-123469188 on Build GRCh38
rs566124978 Chr.7: 123469217-123469217 on Build GRCh38
rs749993180 Chr.7: 123469181-123469181 on Build GRCh38
rs1025830057 Chr.7: 123469159-123469160 on Build GRCh38
rs752899092 Chr.7: 123469228-123469228 on Build GRCh38
rs6972588 Chr.7: 123469177-123469177 on Build GRCh38
rs149235052 Chr.7: 123469238-123469238 on Build GRCh38
rs747995908 Chr.7: 123469193-123469193 on Build GRCh38
rs753319436 Chr.7: 123469256-123469256 on Build GRCh38
rs981094369 Chr.7: 123469108-123469108 on Build GRCh38
rs796209630 Chr.7: 123469137-123469137 on Build GRCh38
rs764441640 Chr.7: 123469231-123469231 on Build GRCh38
rs1051773317 Chr.7: 123469232-123469232 on Build GRCh38
rs973673741 Chr.7: 123469198-123469198 on Build GRCh38
rs570364945 Chr.7: 123469134-123469134 on Build GRCh38
rs74488511 Chr.7: 123469222-123469222 on Build GRCh38
rs748775171 Chr.7: 123469168-123469168 on Build GRCh38
rs781689082 Chr.7: 123469253-123469253 on Build GRCh38
rs1034894053 Chr.7: 123469133-123469133 on Build GRCh38
rs778756391 Chr.7: 123469265-123469265 on Build GRCh38
rs57559552 Chr.7: 123469135-123469135 on Build GRCh38
rs765471285 Chr.7: 123469225-123469225 on Build GRCh38
rs757720186 Chr.7: 123469246-123469246 on Build GRCh38
rs755718629 Chr.7: 123469163-123469163 on Build GRCh38
rs549579150 Chr.7: 123469211-123469211 on Build GRCh38
rs754506374 Chr.7: 123469257-123469257 on Build GRCh38
rs778668638 Chr.7: 123469192-123469192 on Build GRCh38
rs960808951 Chr.7: 123469144-123469144 on Build GRCh38
rs779436921 Chr.7: 123469166-123469166 on Build GRCh38

View Full Product Details