Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57216961-57217198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXPH4
Entrez Gene ID
11247
Gene Name
neurexophilin 4
Gene Aliases
NPH4
UniGene
Hs.745023
Gene Chromosome Location
Chr.12: 57216795-57226449 on Build GRCh38
Transcripts
NM_007224.3 XM_017018747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6354633 c.53G>C p.R18T Chr.12: 57217022-57217022 on Build GRCh38
7212024 c.30G>A p.L10L Chr.12: 57216999-57216999 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761470024 Chr.12: 57217011-57217011 on Build GRCh38
rs758415884 Chr.12: 57217020-57217020 on Build GRCh38
rs971327847 Chr.12: 57217098-57217098 on Build GRCh38
rs763519085 Chr.12: 57216983-57216983 on Build GRCh38
rs1032782758 Chr.12: 57217057-57217057 on Build GRCh38
rs774951798 Chr.12: 57217073-57217073 on Build GRCh38
rs760230527 Chr.12: 57217078-57217078 on Build GRCh38
rs769985754 Chr.12: 57217044-57217044 on Build GRCh38
rs771427025 Chr.12: 57216994-57216994 on Build GRCh38
rs899640726 Chr.12: 57217072-57217072 on Build GRCh38
rs751727034 Chr.12: 57217032-57217032 on Build GRCh38
rs775039840 Chr.12: 57216996-57216996 on Build GRCh38
rs371735726 Chr.12: 57217027-57217027 on Build GRCh38
rs929934613 Chr.12: 57217067-57217067 on Build GRCh38
rs762841339 Chr.12: 57217121-57217121 on Build GRCh38
rs748410710 Chr.12: 57217043-57217043 on Build GRCh38
rs375295780 Chr.12: 57217015-57217015 on Build GRCh38
rs749745839 Chr.12: 57217052-57217052 on Build GRCh38
rs958582457 Chr.12: 57217130-57217130 on Build GRCh38
rs972766455 Chr.12: 57217153-57217153 on Build GRCh38
rs781513968 Chr.12: 57217042-57217042 on Build GRCh38
rs991271338 Chr.12: 57217140-57217141 on Build GRCh38
rs548031803 Chr.12: 57217179-57217179 on Build GRCh38
rs941334290 Chr.12: 57217144-57217144 on Build GRCh38
rs550097331 Chr.12: 57217004-57217004 on Build GRCh38
rs771536207 Chr.12: 57217063-57217063 on Build GRCh38
rs1004833007 Chr.12: 57217147-57217147 on Build GRCh38
rs757498257 Chr.12: 57217042-57217043 on Build GRCh38
rs1034626189 Chr.12: 57217123-57217123 on Build GRCh38
rs753611581 Chr.12: 57217010-57217010 on Build GRCh38
rs1009869146 Chr.12: 57217081-57217081 on Build GRCh38
rs369965960 Chr.12: 57217018-57217018 on Build GRCh38
rs763777178 Chr.12: 57217009-57217009 on Build GRCh38
rs778212253 Chr.12: 57217048-57217048 on Build GRCh38
rs909975661 Chr.12: 57217141-57217141 on Build GRCh38
rs755226727 Chr.12: 57217040-57217040 on Build GRCh38
rs996677386 Chr.12: 57217088-57217088 on Build GRCh38
rs565453827 Chr.12: 57216979-57216979 on Build GRCh38
rs1051762686 Chr.12: 57217099-57217099 on Build GRCh38
rs145729803 Chr.12: 57217103-57217103 on Build GRCh38

View Full Product Details