Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1108692-1108939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SBNO2
Entrez Gene ID
22904
Gene Name
strawberry notch homolog 2
Gene Aliases
KIAA0963, SNO, STNO
UniGene
Hs.408708
Gene Chromosome Location
Chr.19: 1107634-1174283 on Build GRCh38
Transcripts
NM_001100122.1 NM_014963.2 XM_011527804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6772240 c.3349G>A p.V1117M Chr.19: 1108875-1108875 on Build GRCh38
1189746 c.3562G>A p.A1188T Chr.19: 1108887-1108887 on Build GRCh38
6381832 c.3539G>A p.R1180Q Chr.19: 1108910-1108910 on Build GRCh38
4490015 c.3567C>T p.S1189S Chr.19: 1108828-1108828 on Build GRCh38
2813453 c.3558C>T p.C1186C Chr.19: 1108891-1108891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752295232 Chr.19: 1108730-1108730 on Build GRCh38
rs573603595 Chr.19: 1108714-1108714 on Build GRCh38
rs370717411 Chr.19: 1108789-1108789 on Build GRCh38
rs767657865 Chr.19: 1108884-1108884 on Build GRCh38
rs781549839 Chr.19: 1108838-1108839 on Build GRCh38
rs371421677 Chr.19: 1108866-1108866 on Build GRCh38
rs779575057 Chr.19: 1108915-1108915 on Build GRCh38
rs749693484 Chr.19: 1108899-1108899 on Build GRCh38
rs753639305 Chr.19: 1108798-1108798 on Build GRCh38
rs201177605 Chr.19: 1108855-1108855 on Build GRCh38
rs778267107 Chr.19: 1108843-1108843 on Build GRCh38
rs751674389 Chr.19: 1108828-1108828 on Build GRCh38
rs761628064 Chr.19: 1108865-1108865 on Build GRCh38
rs775124011 Chr.19: 1108792-1108792 on Build GRCh38
rs780249690 Chr.19: 1108751-1108751 on Build GRCh38
rs757656438 Chr.19: 1108902-1108902 on Build GRCh38
rs750326734 Chr.19: 1108804-1108804 on Build GRCh38
rs770057414 Chr.19: 1108838-1108838 on Build GRCh38
rs748540310 Chr.19: 1108761-1108761 on Build GRCh38
rs767137863 Chr.19: 1108719-1108719 on Build GRCh38
rs376128681 Chr.19: 1108768-1108768 on Build GRCh38
rs1051667295 Chr.19: 1108858-1108858 on Build GRCh38
rs771517555 Chr.19: 1108847-1108847 on Build GRCh38
rs762118022 Chr.19: 1108816-1108816 on Build GRCh38
rs1047423153 Chr.19: 1108732-1108732 on Build GRCh38
rs749744087 Chr.19: 1108846-1108846 on Build GRCh38
rs760372726 Chr.19: 1108795-1108795 on Build GRCh38
rs757222589 Chr.19: 1108741-1108741 on Build GRCh38
rs765560127 Chr.19: 1108728-1108728 on Build GRCh38
rs751586508 Chr.19: 1108882-1108882 on Build GRCh38
rs185699102 Chr.19: 1108756-1108756 on Build GRCh38
rs753726220 Chr.19: 1108735-1108735 on Build GRCh38
rs760284883 Chr.19: 1108852-1108852 on Build GRCh38
rs761715996 Chr.19: 1108801-1108801 on Build GRCh38
rs765769914 Chr.19: 1108710-1108710 on Build GRCh38
rs763967934 Chr.19: 1108733-1108733 on Build GRCh38
rs781500675 Chr.19: 1108757-1108757 on Build GRCh38
rs891745113 Chr.19: 1108861-1108861 on Build GRCh38
rs979831464 Chr.19: 1108735-1108736 on Build GRCh38
rs766367199 Chr.19: 1108879-1108879 on Build GRCh38
rs112869190 Chr.19: 1108881-1108881 on Build GRCh38
rs368678431 Chr.19: 1108920-1108920 on Build GRCh38
rs564379739 Chr.19: 1108755-1108755 on Build GRCh38
rs777977913 Chr.19: 1108893-1108893 on Build GRCh38
rs1034077379 Chr.19: 1108914-1108914 on Build GRCh38
rs200551019 Chr.19: 1108856-1108856 on Build GRCh38
rs373135662 Chr.19: 1108743-1108743 on Build GRCh38
rs761001783 Chr.19: 1108717-1108717 on Build GRCh38
rs759018791 Chr.19: 1108785-1108785 on Build GRCh38
rs757700276 Chr.19: 1108828-1108830 on Build GRCh38
rs374174588 Chr.19: 1108867-1108867 on Build GRCh38
rs750461485 Chr.19: 1108746-1108746 on Build GRCh38
rs752909677 Chr.19: 1108888-1108888 on Build GRCh38
rs559953905 Chr.19: 1108740-1108740 on Build GRCh38
rs758988648 Chr.19: 1108716-1108716 on Build GRCh38
rs760470589 Chr.19: 1108731-1108731 on Build GRCh38
rs762773680 Chr.19: 1108876-1108876 on Build GRCh38
rs968471754 Chr.19: 1108718-1108718 on Build GRCh38
rs117088896 Chr.19: 1108747-1108747 on Build GRCh38
rs771478042 Chr.19: 1108786-1108786 on Build GRCh38
rs374121379 Chr.19: 1108832-1108832 on Build GRCh38
rs567360270 Chr.19: 1108863-1108863 on Build GRCh38
rs747280041 Chr.19: 1108769-1108770 on Build GRCh38
rs1000042851 Chr.19: 1108824-1108824 on Build GRCh38
rs763871814 Chr.19: 1108797-1108797 on Build GRCh38
rs147525230 Chr.19: 1108766-1108766 on Build GRCh38
rs775100641 Chr.19: 1108851-1108851 on Build GRCh38
rs756416290 Chr.19: 1108891-1108891 on Build GRCh38
rs765157269 Chr.19: 1108802-1108802 on Build GRCh38
rs748452346 Chr.19: 1108834-1108834 on Build GRCh38
rs909891603 Chr.19: 1108796-1108796 on Build GRCh38
rs758422228 Chr.19: 1108809-1108809 on Build GRCh38

View Full Product Details