Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45036120-45036345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSWIM5
Entrez Gene ID
57643
Gene Name
zinc finger SWIM-type containing 5
Gene Aliases
-
UniGene
Hs.656613
Gene Chromosome Location
Chr.1: 45016399-45206586 on Build GRCh38
Transcripts
NM_020883.1 XM_011541861.2 XM_017001913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6444863 c.1910G>A p.R637K Chr.1: 45036284-45036284 on Build GRCh38
4569754 c.1991T>C p.V664A Chr.1: 45036203-45036203 on Build GRCh38
5846833 c.2072A>C p.Q691P Chr.1: 45036122-45036122 on Build GRCh38
3944129 c.1945G>T p.G649C Chr.1: 45036249-45036249 on Build GRCh38
136131 c.2059C>T p.R687C Chr.1: 45036135-45036135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375787755 Chr.1: 45036232-45036232 on Build GRCh38
rs1049510801 Chr.1: 45036248-45036248 on Build GRCh38
rs375562117 Chr.1: 45036142-45036142 on Build GRCh38
rs771729124 Chr.1: 45036252-45036252 on Build GRCh38
rs781497326 Chr.1: 45036195-45036195 on Build GRCh38
rs766989932 Chr.1: 45036288-45036288 on Build GRCh38
rs749219790 Chr.1: 45036218-45036218 on Build GRCh38
rs924717509 Chr.1: 45036284-45036284 on Build GRCh38
rs759921143 Chr.1: 45036296-45036296 on Build GRCh38
rs760265989 Chr.1: 45036261-45036261 on Build GRCh38
rs138503473 Chr.1: 45036291-45036291 on Build GRCh38
rs938645380 Chr.1: 45036313-45036313 on Build GRCh38
rs796208296 Chr.1: 45036266-45036266 on Build GRCh38
rs189694612 Chr.1: 45036257-45036257 on Build GRCh38
rs763792864 Chr.1: 45036157-45036157 on Build GRCh38
rs1048415449 Chr.1: 45036160-45036160 on Build GRCh38
rs560988958 Chr.1: 45036312-45036312 on Build GRCh38
rs770875965 Chr.1: 45036231-45036231 on Build GRCh38
rs765838728 Chr.1: 45036276-45036279 on Build GRCh38
rs201715726 Chr.1: 45036274-45036274 on Build GRCh38
rs767873863 Chr.1: 45036297-45036297 on Build GRCh38
rs372440667 Chr.1: 45036194-45036194 on Build GRCh38
rs1041667126 Chr.1: 45036219-45036219 on Build GRCh38
rs901353242 Chr.1: 45036222-45036222 on Build GRCh38
rs753360092 Chr.1: 45036162-45036162 on Build GRCh38
rs867300991 Chr.1: 45036234-45036234 on Build GRCh38
rs748251288 Chr.1: 45036199-45036199 on Build GRCh38
rs373322583 Chr.1: 45036278-45036278 on Build GRCh38
rs199884737 Chr.1: 45036154-45036154 on Build GRCh38
rs975607205 Chr.1: 45036267-45036267 on Build GRCh38
rs776184222 Chr.1: 45036276-45036276 on Build GRCh38
rs760336715 Chr.1: 45036141-45036141 on Build GRCh38
rs745654748 Chr.1: 45036238-45036238 on Build GRCh38
rs201379465 Chr.1: 45036279-45036279 on Build GRCh38
rs756096101 Chr.1: 45036216-45036216 on Build GRCh38
rs200925379 Chr.1: 45036153-45036153 on Build GRCh38

View Full Product Details