Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40222579-40222824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RLF
Entrez Gene ID
6018
Gene Name
rearranged L-myc fusion
Gene Aliases
ZN-15L, ZNF292L
UniGene
Hs.205627
Gene Chromosome Location
Chr.1: 40161369-40240921 on Build GRCh38
Transcripts
NM_012421.3 XM_017002016.1 XM_017002017.1 XM_017002018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
909393 c.878A>G p.N293S Chr.1: 40222641-40222641 on Build GRCh38
6379870 c.916C>A p.Q306K Chr.1: 40222679-40222679 on Build GRCh38
1667462 c.937T>G p.Y313D Chr.1: 40222700-40222700 on Build GRCh38
6414665 c.825C>G p.D275E Chr.1: 40222588-40222588 on Build GRCh38
5538263 c.863C>T p.S288F Chr.1: 40222626-40222626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773770639 Chr.1: 40222732-40222732 on Build GRCh38
rs780330321 Chr.1: 40222607-40222607 on Build GRCh38
rs759015900 Chr.1: 40222755-40222755 on Build GRCh38
rs769581201 Chr.1: 40222663-40222663 on Build GRCh38
rs747042111 Chr.1: 40222735-40222736 on Build GRCh38
rs767025065 Chr.1: 40222758-40222758 on Build GRCh38
rs1016840003 Chr.1: 40222648-40222648 on Build GRCh38
rs758329060 Chr.1: 40222771-40222771 on Build GRCh38
rs1029296618 Chr.1: 40222767-40222767 on Build GRCh38
rs955024788 Chr.1: 40222788-40222788 on Build GRCh38
rs773125277 Chr.1: 40222699-40222699 on Build GRCh38
rs781497751 Chr.1: 40222641-40222641 on Build GRCh38
rs749277300 Chr.1: 40222704-40222704 on Build GRCh38
rs759111051 Chr.1: 40222736-40222736 on Build GRCh38
rs770402774 Chr.1: 40222718-40222718 on Build GRCh38
rs375968085 Chr.1: 40222750-40222750 on Build GRCh38
rs768718921 Chr.1: 40222632-40222632 on Build GRCh38
rs958909405 Chr.1: 40222734-40222734 on Build GRCh38
rs758539726 Chr.1: 40222605-40222605 on Build GRCh38
rs775216300 Chr.1: 40222745-40222745 on Build GRCh38
rs996518147 Chr.1: 40222766-40222766 on Build GRCh38
rs991337598 Chr.1: 40222735-40222738 on Build GRCh38
rs181906563 Chr.1: 40222645-40222645 on Build GRCh38
rs767021882 Chr.1: 40222742-40222742 on Build GRCh38
rs369227999 Chr.1: 40222753-40222753 on Build GRCh38
rs747839448 Chr.1: 40222662-40222662 on Build GRCh38
rs747226002 Chr.1: 40222619-40222619 on Build GRCh38

View Full Product Details