Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66057167-66057440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SF3B2
Entrez Gene ID
10992
Gene Name
splicing factor 3b subunit 2
Gene Aliases
Cus1, SAP145, SF3B145, SF3b1, SF3b150
UniGene
Hs.406423
Gene Chromosome Location
Chr.11: 66051772-66068911 on Build GRCh38
Transcripts
NM_006842.2 XM_005273726.3 XM_011544740.2 XM_017017144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6776260 c.734G>A p.R245Q Chr.11: 66057332-66057332 on Build GRCh38
415810 c.676C>G p.P226A Chr.11: 66057274-66057274 on Build GRCh38
6070021 c.704C>T p.T235I Chr.11: 66057302-66057302 on Build GRCh38
4035627 c.777+2T>C p.? Chr.11: 66057377-66057377 on Build GRCh38
6519257 c.714C>A p.P238P Chr.11: 66057312-66057312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764085353 Chr.11: 66057405-66057406 on Build GRCh38
rs988727736 Chr.11: 66057265-66057265 on Build GRCh38
rs759761058 Chr.11: 66057378-66057378 on Build GRCh38
rs762987980 Chr.11: 66057394-66057396 on Build GRCh38
rs767486871 Chr.11: 66057274-66057282 on Build GRCh38
rs757168089 Chr.11: 66057299-66057299 on Build GRCh38
rs750399098 Chr.11: 66057307-66057307 on Build GRCh38
rs779881778 Chr.11: 66057321-66057321 on Build GRCh38
rs761319326 Chr.11: 66057347-66057347 on Build GRCh38
rs377149955 Chr.11: 66057222-66057222 on Build GRCh38
rs77713633 Chr.11: 66057350-66057350 on Build GRCh38
rs979032461 Chr.11: 66057251-66057251 on Build GRCh38
rs372204235 Chr.11: 66057266-66057266 on Build GRCh38
rs950808255 Chr.11: 66057340-66057340 on Build GRCh38
rs912031297 Chr.11: 66057387-66057387 on Build GRCh38
rs773749878 Chr.11: 66057235-66057235 on Build GRCh38
rs968573642 Chr.11: 66057393-66057393 on Build GRCh38
rs765399704 Chr.11: 66057394-66057394 on Build GRCh38
rs201649157 Chr.11: 66057328-66057328 on Build GRCh38
rs886612846 Chr.11: 66057332-66057332 on Build GRCh38
rs930396745 Chr.11: 66057318-66057318 on Build GRCh38
rs754095623 Chr.11: 66057280-66057280 on Build GRCh38
rs372263537 Chr.11: 66057220-66057220 on Build GRCh38
rs369292605 Chr.11: 66057256-66057256 on Build GRCh38
rs1028983292 Chr.11: 66057360-66057360 on Build GRCh38
rs374892362 Chr.11: 66057313-66057313 on Build GRCh38
rs902080717 Chr.11: 66057338-66057338 on Build GRCh38
rs745683375 Chr.11: 66057215-66057215 on Build GRCh38
rs772490799 Chr.11: 66057239-66057239 on Build GRCh38
rs771360430 Chr.11: 66057348-66057348 on Build GRCh38
rs949710338 Chr.11: 66057286-66057286 on Build GRCh38
rs747635710 Chr.11: 66057331-66057331 on Build GRCh38
rs925802932 Chr.11: 66057253-66057253 on Build GRCh38
rs747292956 Chr.11: 66057393-66057393 on Build GRCh38
rs768199249 Chr.11: 66057234-66057234 on Build GRCh38
rs749008352 Chr.11: 66057225-66057225 on Build GRCh38
rs762911141 Chr.11: 66057408-66057408 on Build GRCh38
rs753011114 Chr.11: 66057396-66057396 on Build GRCh38
rs762280409 Chr.11: 66057337-66057337 on Build GRCh38
rs766894011 Chr.11: 66057276-66057276 on Build GRCh38
rs771761232 Chr.11: 66057335-66057335 on Build GRCh38
rs1044043353 Chr.11: 66057291-66057291 on Build GRCh38
rs759688428 Chr.11: 66057283-66057283 on Build GRCh38
rs200330809 Chr.11: 66057294-66057294 on Build GRCh38
rs575508105 Chr.11: 66057257-66057257 on Build GRCh38
rs369029996 Chr.11: 66057314-66057314 on Build GRCh38
rs769626118 Chr.11: 66057216-66057216 on Build GRCh38
rs762315511 Chr.11: 66057238-66057238 on Build GRCh38
rs752749258 Chr.11: 66057298-66057298 on Build GRCh38
rs187615832 Chr.11: 66057301-66057301 on Build GRCh38

View Full Product Details