Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2997799-2998068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLE2
Entrez Gene ID
7089
Gene Name
transducin like enhancer of split 2
Gene Aliases
ESG, ESG2, GRG2
UniGene
Hs.332173
Gene Chromosome Location
Chr.19: 2997638-3047635 on Build GRCh38
Transcripts
NM_001144761.1 NM_001144762.1 NM_001300846.1 NM_003260.4 XM_006722864.2 XM_011528230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
994374 c.2195G>T p.G732V Chr.19: 2997885-2997885 on Build GRCh38
6557239 c.2178C>A p.Y726* Chr.19: 2997902-2997902 on Build GRCh38
4154152 c.2202G>T p.K734N Chr.19: 2997878-2997878 on Build GRCh38
3822612 c.2155G>C p.D719H Chr.19: 2997925-2997925 on Build GRCh38
5722780 c.2141C>T p.S714L Chr.19: 2997939-2997939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773949175 Chr.19: 2997835-2997835 on Build GRCh38
rs755096507 Chr.19: 2997918-2997918 on Build GRCh38
rs941362961 Chr.19: 2998006-2998006 on Build GRCh38
rs765163824 Chr.19: 2997898-2997898 on Build GRCh38
rs762105247 Chr.19: 2997827-2997827 on Build GRCh38
rs761890014 Chr.19: 2997963-2997963 on Build GRCh38
rs373620175 Chr.19: 2997941-2997941 on Build GRCh38
rs1050260073 Chr.19: 2998044-2998044 on Build GRCh38
rs111313851 Chr.19: 2997887-2997887 on Build GRCh38
rs771690849 Chr.19: 2997951-2997951 on Build GRCh38
rs889039790 Chr.19: 2997883-2997883 on Build GRCh38
rs1038887385 Chr.19: 2997834-2997834 on Build GRCh38
rs974073132 Chr.19: 2998013-2998013 on Build GRCh38
rs940047573 Chr.19: 2998008-2998008 on Build GRCh38
rs759230997 Chr.19: 2997841-2997841 on Build GRCh38
rs416799 Chr.19: 2997978-2997978 on Build GRCh38
rs200910426 Chr.19: 2997953-2997953 on Build GRCh38
rs371891336 Chr.19: 2997945-2997945 on Build GRCh38
rs762011716 Chr.19: 2997906-2997908 on Build GRCh38
rs1018870033 Chr.19: 2997888-2997888 on Build GRCh38
rs897364219 Chr.19: 2998014-2998014 on Build GRCh38
rs750309415 Chr.19: 2997968-2997968 on Build GRCh38
rs370834728 Chr.19: 2997823-2997823 on Build GRCh38
rs774685874 Chr.19: 2997834-2997835 on Build GRCh38
rs775214263 Chr.19: 2997952-2997952 on Build GRCh38
rs767035393 Chr.19: 2997848-2997848 on Build GRCh38
rs774924243 Chr.19: 2997857-2997857 on Build GRCh38
rs981699962 Chr.19: 2997986-2997986 on Build GRCh38
rs374054297 Chr.19: 2997824-2997824 on Build GRCh38
rs770163177 Chr.19: 2997828-2997828 on Build GRCh38
rs898567589 Chr.19: 2997845-2997845 on Build GRCh38
rs776670491 Chr.19: 2997962-2997962 on Build GRCh38
rs11150 Chr.19: 2997899-2997899 on Build GRCh38
rs932714628 Chr.19: 2998033-2998033 on Build GRCh38
rs780176029 Chr.19: 2997904-2997904 on Build GRCh38
rs372279902 Chr.19: 2997934-2997934 on Build GRCh38
rs201815984 Chr.19: 2997961-2997961 on Build GRCh38
rs199863291 Chr.19: 2997944-2997944 on Build GRCh38
rs756675212 Chr.19: 2997996-2997996 on Build GRCh38
rs1024429542 Chr.19: 2997840-2997840 on Build GRCh38
rs746991756 Chr.19: 2997915-2997915 on Build GRCh38
rs113553089 Chr.19: 2998032-2998032 on Build GRCh38
rs778108986 Chr.19: 2997997-2997997 on Build GRCh38
rs199703619 Chr.19: 2997869-2997869 on Build GRCh38
rs950308872 Chr.19: 2998025-2998025 on Build GRCh38
rs763682574 Chr.19: 2997958-2997958 on Build GRCh38
rs200647783 Chr.19: 2997868-2997868 on Build GRCh38
rs201001610 Chr.19: 2997972-2997972 on Build GRCh38
rs765110498 Chr.19: 2997964-2997964 on Build GRCh38
rs374457134 Chr.19: 2997935-2997935 on Build GRCh38
rs748636424 Chr.19: 2997981-2997981 on Build GRCh38
rs781625226 Chr.19: 2997923-2997923 on Build GRCh38
rs551037753 Chr.19: 2998049-2998049 on Build GRCh38
rs539047567 Chr.19: 2998012-2998012 on Build GRCh38
rs375354757 Chr.19: 2997970-2997970 on Build GRCh38
rs781286645 Chr.19: 2997980-2997980 on Build GRCh38
rs772475466 Chr.19: 2998003-2998004 on Build GRCh38
rs749551472 Chr.19: 2998005-2998005 on Build GRCh38

View Full Product Details