Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111931556-111931829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM116
Entrez Gene ID
89894
Gene Name
transmembrane protein 116
Gene Aliases
-
UniGene
Hs.506815
Gene Chromosome Location
Chr.12: 111894936-112013219 on Build GRCh38
Transcripts
NM_001193453.1 NM_001193531.1 NM_001294314.1 NM_138341.2 NR_122119.1 XM_006719678.3 XM_011538946.1 XM_011538947.1 XM_011538948.2 XM_011538949.2 XM_011538950.1 XM_011538951.2 XM_011538953.1 XM_011538955.1 XM_011538957.1 XM_011538958.1 XM_017020187.1 XM_017020188.1 XM_017020189.1 XM_017020190.1 XM_017020191.1 XM_017020192.1 XM_017020193.1 XM_017020194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4871125 c.864G>A p.Q288Q Chr.12: 111931771-111931771 on Build GRCh38
6194667 c.826C>G p.Q276E Chr.12: 111931809-111931809 on Build GRCh38
935120 c.588G>A p.Q196Q Chr.12: 111931771-111931771 on Build GRCh38
223597 c.636_637CC>TA p.T212>? Chr.12: 111931722-111931723 on Build GRCh38
4948037 c.860C>T p.T287M Chr.12: 111931775-111931775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750838586 Chr.12: 111931724-111931724 on Build GRCh38
rs773061292 Chr.12: 111931740-111931740 on Build GRCh38
rs559301794 Chr.12: 111931640-111931640 on Build GRCh38
rs375840968 Chr.12: 111931739-111931739 on Build GRCh38
rs759014592 Chr.12: 111931775-111931775 on Build GRCh38
rs765693215 Chr.12: 111931722-111931722 on Build GRCh38
rs963846580 Chr.12: 111931673-111931673 on Build GRCh38
rs773007569 Chr.12: 111931685-111931685 on Build GRCh38
rs757556503 Chr.12: 111931719-111931719 on Build GRCh38
rs139617530 Chr.12: 111931786-111931786 on Build GRCh38
rs117800485 Chr.12: 111931744-111931744 on Build GRCh38
rs780242241 Chr.12: 111931729-111931729 on Build GRCh38
rs369338579 Chr.12: 111931801-111931801 on Build GRCh38
rs371845657 Chr.12: 111931736-111931736 on Build GRCh38
rs771895695 Chr.12: 111931799-111931801 on Build GRCh38
rs1042870300 Chr.12: 111931583-111931583 on Build GRCh38
rs776521476 Chr.12: 111931659-111931659 on Build GRCh38
rs770318114 Chr.12: 111931710-111931710 on Build GRCh38
rs150575127 Chr.12: 111931756-111931756 on Build GRCh38
rs147550432 Chr.12: 111931701-111931701 on Build GRCh38
rs199679612 Chr.12: 111931783-111931783 on Build GRCh38
rs774321068 Chr.12: 111931761-111931761 on Build GRCh38
rs769672878 Chr.12: 111931677-111931677 on Build GRCh38
rs748252807 Chr.12: 111931696-111931696 on Build GRCh38
rs201605168 Chr.12: 111931708-111931708 on Build GRCh38
rs747161836 Chr.12: 111931648-111931648 on Build GRCh38
rs375123049 Chr.12: 111931737-111931737 on Build GRCh38
rs370592954 Chr.12: 111931774-111931774 on Build GRCh38
rs781491775 Chr.12: 111931688-111931688 on Build GRCh38
rs767079388 Chr.12: 111931780-111931780 on Build GRCh38
rs746456861 Chr.12: 111931622-111931622 on Build GRCh38
rs760390393 Chr.12: 111931785-111931785 on Build GRCh38
rs752432882 Chr.12: 111931711-111931711 on Build GRCh38
rs541050668 Chr.12: 111931590-111931590 on Build GRCh38
rs146203211 Chr.12: 111931660-111931660 on Build GRCh38
rs530248992 Chr.12: 111931658-111931658 on Build GRCh38
rs372438649 Chr.12: 111931613-111931613 on Build GRCh38
rs758979269 Chr.12: 111931725-111931725 on Build GRCh38
rs750831853 Chr.12: 111931787-111931787 on Build GRCh38
rs113902987 Chr.12: 111931772-111931772 on Build GRCh38
rs1016390637 Chr.12: 111931669-111931669 on Build GRCh38
rs765804760 Chr.12: 111931694-111931694 on Build GRCh38
rs200298981 Chr.12: 111931641-111931641 on Build GRCh38
rs148439044 Chr.12: 111931675-111931675 on Build GRCh38
rs747112616 Chr.12: 111931734-111931734 on Build GRCh38
rs141724043 Chr.12: 111931707-111931707 on Build GRCh38
rs757687891 Chr.12: 111931621-111931621 on Build GRCh38
rs144475691 Chr.12: 111931790-111931790 on Build GRCh38

View Full Product Details