Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76567360-76567623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST6GALNAC2
Entrez Gene ID
10610
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 2
Gene Aliases
SAITL1, SIAT7, SIAT7B, SIATL1, ST6GalNAII, STHM
UniGene
Hs.592105
Gene Chromosome Location
Chr.17: 76565379-76586066 on Build GRCh38
Transcripts
NM_006456.2 XM_005256954.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
473412 c.876G>C p.L292F Chr.17: 76567534-76567534 on Build GRCh38
1610816 c.879T>C p.I293I Chr.17: 76567531-76567531 on Build GRCh38
6791165 c.950G>A p.C317Y Chr.17: 76567460-76567460 on Build GRCh38
1386234 c.891delT p.F297fs*15 Chr.17: 76567519-76567519 on Build GRCh38
291085 c.949T>C p.C317R Chr.17: 76567461-76567461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772702249 Chr.17: 76567565-76567566 on Build GRCh38
rs753508965 Chr.17: 76567536-76567536 on Build GRCh38
rs909240803 Chr.17: 76567394-76567394 on Build GRCh38
rs147183255 Chr.17: 76567420-76567420 on Build GRCh38
rs772012009 Chr.17: 76567596-76567596 on Build GRCh38
rs200554513 Chr.17: 76567444-76567445 on Build GRCh38
rs371194897 Chr.17: 76567421-76567421 on Build GRCh38
rs770119908 Chr.17: 76567452-76567452 on Build GRCh38
rs750751657 Chr.17: 76567422-76567422 on Build GRCh38
rs1009875910 Chr.17: 76567484-76567484 on Build GRCh38
rs567557862 Chr.17: 76567570-76567570 on Build GRCh38
rs770568660 Chr.17: 76567580-76567580 on Build GRCh38
rs773610483 Chr.17: 76567453-76567453 on Build GRCh38
rs745694326 Chr.17: 76567587-76567587 on Build GRCh38
rs1019879242 Chr.17: 76567531-76567534 on Build GRCh38
rs868787657 Chr.17: 76567549-76567549 on Build GRCh38
rs201476012 Chr.17: 76567458-76567458 on Build GRCh38
rs551933129 Chr.17: 76567494-76567494 on Build GRCh38
rs1033821217 Chr.17: 76567488-76567488 on Build GRCh38
rs752795328 Chr.17: 76567552-76567552 on Build GRCh38
rs754946142 Chr.17: 76567438-76567438 on Build GRCh38
rs781777112 Chr.17: 76567545-76567545 on Build GRCh38
rs555400646 Chr.17: 76567495-76567495 on Build GRCh38
rs751935503 Chr.17: 76567541-76567541 on Build GRCh38
rs758791478 Chr.17: 76567428-76567428 on Build GRCh38
rs749054317 Chr.17: 76567454-76567454 on Build GRCh38
rs933376855 Chr.17: 76567416-76567416 on Build GRCh38
rs758924959 Chr.17: 76567412-76567412 on Build GRCh38
rs763693330 Chr.17: 76567505-76567505 on Build GRCh38
rs774954247 Chr.17: 76567478-76567478 on Build GRCh38
rs755497273 Chr.17: 76567544-76567544 on Build GRCh38
rs2241918 Chr.17: 76567571-76567571 on Build GRCh38
rs771974773 Chr.17: 76567467-76567467 on Build GRCh38
rs767072166 Chr.17: 76567418-76567418 on Build GRCh38
rs759526325 Chr.17: 76567459-76567459 on Build GRCh38
rs1032601982 Chr.17: 76567477-76567477 on Build GRCh38
rs766198446 Chr.17: 76567407-76567407 on Build GRCh38
rs772760001 Chr.17: 76567410-76567410 on Build GRCh38
rs765388230 Chr.17: 76567530-76567530 on Build GRCh38
rs747562975 Chr.17: 76567435-76567435 on Build GRCh38
rs959811624 Chr.17: 76567490-76567490 on Build GRCh38
rs200117442 Chr.17: 76567539-76567539 on Build GRCh38
rs937280733 Chr.17: 76567383-76567384 on Build GRCh38
rs756295365 Chr.17: 76567566-76567566 on Build GRCh38
rs143735856 Chr.17: 76567399-76567399 on Build GRCh38
rs549567808 Chr.17: 76567392-76567392 on Build GRCh38
rs780373837 Chr.17: 76567434-76567434 on Build GRCh38
rs748296298 Chr.17: 76567446-76567446 on Build GRCh38
rs753930089 Chr.17: 76567398-76567398 on Build GRCh38
rs781328035 Chr.17: 76567443-76567443 on Build GRCh38
rs1005990508 Chr.17: 76567424-76567424 on Build GRCh38
rs778768290 Chr.17: 76567586-76567586 on Build GRCh38

View Full Product Details