Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 48233578-48233847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC5A9
Entrez Gene ID
200010
Gene Name
solute carrier family 5 member 9
Gene Aliases
SGLT4
UniGene
Hs.37890
Gene Chromosome Location
Chr.1: 48221113-48248644 on Build GRCh38
Transcripts
NM_001011547.2 NM_001135181.1 XM_011540924.2 XM_011540925.2 XM_011540926.2 XM_011540927.1 XM_011540928.1 XM_011540929.2 XM_017000558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
910439 c.1124T>C p.V375A Chr.1: 48233670-48233670 on Build GRCh38
4521692 c.1125G>A p.V375V Chr.1: 48233671-48233671 on Build GRCh38
7091379 c.1145G>T p.R382I Chr.1: 48233691-48233691 on Build GRCh38
4521454 c.1111G>A p.E371K Chr.1: 48233657-48233657 on Build GRCh38
3865731 c.1155G>A p.G385G Chr.1: 48233701-48233701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781756895 Chr.1: 48233741-48233741 on Build GRCh38
rs1015215514 Chr.1: 48233826-48233826 on Build GRCh38
rs952945305 Chr.1: 48233649-48233649 on Build GRCh38
rs376023192 Chr.1: 48233669-48233669 on Build GRCh38
rs139285576 Chr.1: 48233680-48233680 on Build GRCh38
rs1014091685 Chr.1: 48233759-48233759 on Build GRCh38
rs961943575 Chr.1: 48233625-48233625 on Build GRCh38
rs759472113 Chr.1: 48233788-48233788 on Build GRCh38
rs369126505 Chr.1: 48233757-48233757 on Build GRCh38
rs755912372 Chr.1: 48233691-48233692 on Build GRCh38
rs765258459 Chr.1: 48233791-48233791 on Build GRCh38
rs752560649 Chr.1: 48233653-48233653 on Build GRCh38
rs768225318 Chr.1: 48233750-48233750 on Build GRCh38
rs759668252 Chr.1: 48233681-48233681 on Build GRCh38
rs375673412 Chr.1: 48233617-48233617 on Build GRCh38
rs148677917 Chr.1: 48233675-48233675 on Build GRCh38
rs201757852 Chr.1: 48233668-48233668 on Build GRCh38
rs770282251 Chr.1: 48233656-48233656 on Build GRCh38
rs945575414 Chr.1: 48233733-48233733 on Build GRCh38
rs952997439 Chr.1: 48233824-48233824 on Build GRCh38
rs761573016 Chr.1: 48233659-48233659 on Build GRCh38
rs199879523 Chr.1: 48233694-48233694 on Build GRCh38
rs753044269 Chr.1: 48233608-48233608 on Build GRCh38
rs775567764 Chr.1: 48233811-48233811 on Build GRCh38
rs367990112 Chr.1: 48233771-48233771 on Build GRCh38
rs567259297 Chr.1: 48233657-48233657 on Build GRCh38
rs778574259 Chr.1: 48233753-48233753 on Build GRCh38
rs776490543 Chr.1: 48233786-48233786 on Build GRCh38
rs905375273 Chr.1: 48233806-48233806 on Build GRCh38
rs746884409 Chr.1: 48233781-48233781 on Build GRCh38
rs747585205 Chr.1: 48233623-48233623 on Build GRCh38
rs925883686 Chr.1: 48233730-48233730 on Build GRCh38
rs982440027 Chr.1: 48233653-48233653 on Build GRCh38
rs372099516 Chr.1: 48233624-48233624 on Build GRCh38
rs1004418510 Chr.1: 48233825-48233825 on Build GRCh38
rs1043014320 Chr.1: 48233719-48233719 on Build GRCh38
rs765311609 Chr.1: 48233685-48233685 on Build GRCh38
rs757674996 Chr.1: 48233737-48233737 on Build GRCh38
rs144112204 Chr.1: 48233706-48233706 on Build GRCh38
rs149560761 Chr.1: 48233665-48233665 on Build GRCh38
rs758650324 Chr.1: 48233705-48233705 on Build GRCh38
rs769437039 Chr.1: 48233807-48233807 on Build GRCh38
rs779408147 Chr.1: 48233643-48233643 on Build GRCh38
rs530532743 Chr.1: 48233616-48233616 on Build GRCh38
rs778376175 Chr.1: 48233618-48233618 on Build GRCh38
rs758884138 Chr.1: 48233605-48233605 on Build GRCh38
rs762761457 Chr.1: 48233740-48233740 on Build GRCh38
rs79196143 Chr.1: 48233778-48233778 on Build GRCh38
rs752848191 Chr.1: 48233703-48233703 on Build GRCh38
rs138756482 Chr.1: 48233822-48233822 on Build GRCh38
rs553678770 Chr.1: 48233732-48233732 on Build GRCh38

View Full Product Details