Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96864696-96864967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA4C
Entrez Gene ID
54910
Gene Name
semaphorin 4C
Gene Aliases
M-SEMA-F, SEMACL1, SEMAF, SEMAI
UniGene
Hs.516220
Gene Chromosome Location
Chr.2: 96859736-96870943 on Build GRCh38
Transcripts
NM_017789.4 XM_006712606.3 XM_011511378.2 XM_011511379.2 XM_011511380.1 XM_011511381.1 XM_011511382.2 XM_011511383.1 XM_017004393.1 XM_017004394.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1024012 c.789C>T p.G263G Chr.2: 96864878-96864878 on Build GRCh38
1471353 c.853T>C p.C285R Chr.2: 96864814-96864814 on Build GRCh38
1738669 c.799G>C p.G267R Chr.2: 96864868-96864868 on Build GRCh38
4781678 c.787-9G>A p.? Chr.2: 96864889-96864889 on Build GRCh38
7133106 c.844C>A p.R282R Chr.2: 96864823-96864823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986287419 Chr.2: 96864881-96864881 on Build GRCh38
rs754319720 Chr.2: 96864832-96864832 on Build GRCh38
rs752887211 Chr.2: 96864865-96864865 on Build GRCh38
rs150540972 Chr.2: 96864750-96864750 on Build GRCh38
rs745849511 Chr.2: 96864895-96864895 on Build GRCh38
rs748831563 Chr.2: 96864906-96864906 on Build GRCh38
rs771261568 Chr.2: 96864931-96864931 on Build GRCh38
rs779761560 Chr.2: 96864902-96864902 on Build GRCh38
rs775266807 Chr.2: 96864801-96864801 on Build GRCh38
rs745318332 Chr.2: 96864783-96864783 on Build GRCh38
rs781017776 Chr.2: 96864781-96864781 on Build GRCh38
rs773578797 Chr.2: 96864815-96864815 on Build GRCh38
rs777990010 Chr.2: 96864878-96864878 on Build GRCh38
rs372109047 Chr.2: 96864738-96864738 on Build GRCh38
rs772544739 Chr.2: 96864811-96864811 on Build GRCh38
rs567359236 Chr.2: 96864804-96864804 on Build GRCh38
rs752978770 Chr.2: 96864758-96864758 on Build GRCh38
rs769440981 Chr.2: 96864799-96864799 on Build GRCh38
rs781126171 Chr.2: 96864890-96864890 on Build GRCh38
rs767572658 Chr.2: 96864947-96864947 on Build GRCh38
rs777582579 Chr.2: 96864771-96864771 on Build GRCh38
rs550741731 Chr.2: 96864773-96864773 on Build GRCh38
rs779053245 Chr.2: 96864755-96864755 on Build GRCh38
rs538940437 Chr.2: 96864946-96864946 on Build GRCh38
rs955856957 Chr.2: 96864837-96864837 on Build GRCh38
rs185401533 Chr.2: 96864889-96864889 on Build GRCh38
rs978374370 Chr.2: 96864923-96864923 on Build GRCh38
rs758187273 Chr.2: 96864770-96864770 on Build GRCh38
rs139084624 Chr.2: 96864824-96864824 on Build GRCh38
rs773879115 Chr.2: 96864916-96864916 on Build GRCh38
rs962280833 Chr.2: 96864805-96864805 on Build GRCh38
rs758666759 Chr.2: 96864872-96864872 on Build GRCh38
rs113960365 Chr.2: 96864917-96864917 on Build GRCh38
rs746895608 Chr.2: 96864772-96864772 on Build GRCh38
rs765370801 Chr.2: 96864849-96864849 on Build GRCh38
rs145384778 Chr.2: 96864836-96864836 on Build GRCh38
rs148034324 Chr.2: 96864735-96864735 on Build GRCh38
rs141614070 Chr.2: 96864803-96864803 on Build GRCh38
rs760978241 Chr.2: 96864822-96864822 on Build GRCh38
rs367619112 Chr.2: 96864907-96864907 on Build GRCh38
rs267599502 Chr.2: 96864747-96864747 on Build GRCh38

View Full Product Details